Merck
  • Pericellular matrix of decidua-derived mesenchymal cells: a potent human-derived substrate for the maintenance culture of human ES cells.
Pericellular matrix of decidua-derived mesenchymal cells: a potent human-derived substrate for the maintenance culture of human ES cells.
Developmental dynamics : an official publication of the American Association of Anatomists (2009-04-23)
Tomoko Nagase, Morio Ueno, Michiru Matsumura, Keiko Muguruma, Masatoshi Ohgushi, Naomi Kondo, Daisuke Kanematsu, Yonehiro Kanemura, Yoshiki Sasai
ABSTRACT
In routine culture, human embryonic stem (hES) cells are maintained on either feeder cells or special culture substrates such as Matrigel. However, to expand hES cells for clinical applications, it is desirable to minimize animal-derived materials in the culture for safety reasons. In this report, we show that the pericellular matrix prepared from human decidua-derived mesenchymal cells (PCM-DM) is a potent substrate material that supports the growth and pluripotency of hES cells as efficiently as Matrigel does. This supporting activity of PCM-DM is stable and can be preserved for several months in the refrigerator. PCM-DM-based culture is compatible with non-conditioned commercial defined medium, and with the maintenance of dissociated hES cells in the presence of ROCK inhibitor. Since decidual mesenchymal cells can be prepared and expanded in a large quantity, PCM-DM is a practical human-derived substitute for the animal-derived substrates for use in clinical-grade culture of hES cells.
Sigma-Aldrich
Anti-Vimentin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Fibronectin Frag Antibody, cell attachment fragment, clone 3E3, clone 3E3, Chemicon®, from mouse
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480, clone MC-480, Chemicon®, from mouse
Sigma-Aldrich
Anti-Laminin Antibody, clone 2G6/A2, clone 2G6/A2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse