Skip to Content
Merck
All Photos(1)

Key Documents

MAB13426

Sigma-Aldrich

Anti-MMP-13 Antibody, clone LIPCO IID1

clone LIPCO IID1, Chemicon®, from mouse

Synonym(s):

Collagenase-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

LIPCO IID1, monoclonal

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MMP13(4322)

Specificity

It recognizes proteins of ~60 kDa and ~48 kDa which are identified as pro (latent) and activate forms of matrix metalloproteinase-13 (MMP-13; also known as Collagenase-3). Antibody shows no cross-reactivity with the pro and active forms of other MMPs. Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP-13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22, including fibroblast collagenase (MMP-1), matrilysin (MMP-7), and macrophage metalloelastase (MMP-12), but is more distantly related to genes coding for stromelysin-3 (MMP-11), gelatinase-A (MMP-2), and gelatinase-B (MMP-9), which map outside of this gene cluster.

Cellular Localization:

Cytoplasmic

Immunogen

Rat MMP-13 from BC-1 cells.

Application

Anti-MMP-13 Antibody, clone LIPCO IID1 is an antibody against MMP-13 for use in WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
Western Blotting (Lyons, 1991; Lyons, 1993): 1:200-1:400 at room temperature for 2 hours.

Immunofluorescence

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Analysis Note

Control
POSITIVE CONTROL: Conditioned, serum-free medium from (PMA-treated) rat fibrosarcoma cells or rat keratinocyte CCL-4.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erika Ramos-Tovar et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 49(2), 212-223 (2018-10-20)
The aims of the present study were to investigate the capacity of stevia leaves to prevent experimental cirrhosis induced by chronic administration of carbon tetrachloride (CCl4 ) in rats and to explore the action mechanism involved. Liver cirrhosis was established
Song Ho Chang et al.
Nature communications, 10(1), 1442-1442 (2019-03-31)
Exposure of articular cartilage to excessive mechanical loading is deeply involved in the pathogenesis of osteoarthritis. Here, we identify gremlin-1 as a mechanical loading-inducible factor in chondrocytes, detected at high levels in middle and deep layers of cartilage after cyclic
Kristina Koop et al.
Frontiers in immunology, 14, 1163198-1163198 (2023-05-19)
Fibrostenotic disease is a common complication in Crohn's disease (CD) patients hallmarked by transmural extracellular matrix (ECM) accumulation in the intestinal wall. The prevention and medical therapy of fibrostenotic CD is an unmet high clinical need. Although targeting IL36R signaling
Aya Miyauchi et al.
Scientific reports, 9(1), 15770-15770 (2019-11-02)
Excessive mechanical stress is a major cause of knee osteoarthritis. However, the mechanism by which the mechanical stress begets osteoarthritis development remains elusive. Hydrogen peroxide-inducible clone-5 (Hic-5; TGFβ1i1), a TGF-β inducible focal adhesion adaptor, has previously been reported as a
Xiaochen Li et al.
Frontiers in pharmacology, 12, 690181-690181 (2021-06-22)
Osteoarthritis (OA) is a worldwide degenerative joint disease that seriously impaired the quality of life of patients. OA has been established as a disease with metabolic disorder. Cholesterol 25-hydroxylase (CH25H) was proved to play a key role in cartilage cholesterol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service