Skip to Content
Merck
All Photos(1)

Key Documents

AB1534

Sigma-Aldrich

Anti-Brain Derived Neurotrophic Factor Antibody

serum, Chemicon®

Synonym(s):

brain-derived neurotrophic factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

suitability

not suitable for activity/function inhibition

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

General description

Brain Derived Neurotrophic Factor (BDNF) is a secreted protein that promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. It is a major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability. It is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of this gene is reduced in both Alzheimer’s and Huntington disease patients. BDNF may play a role in the regulation of stress response and in the biology of mood disorders. BDNF exists as monomers or homodimers and binds to TRKB/NTRK2 receptors.

Specificity

Brain Derived Neurotrophic Factor (BDNF). By dot blot the antibody shows less than 0.1% cross reactivity with recombinant human NT3, NT4/5 or mouse NGF.

Immunogen

A 10 amino acid peptide corresponding to the amino-terminal of mouse Brain Derived Neurotrophic Factor (BDNF) coupled to ovalbumin.

Application

Immunohistochemistry:
1:1000-1:2000 dilution of a previous lot was used in immunohistochemistry (see suggested protocol).

ELISA:
1:1000-1:2000 dilution of a previous lot was used in ELISA.

Bioassay:
Does not inhibit BDNF biological activity.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neuroinflammation & Pain
This Anti-Brain Derived Neurotrophic Factor Antibody is validated for use in ELISA, IH, WB for the detection of Brain Derived Neurotrophic Factor.

Quality

Evaluated by Western Blot on mouse brain lysates.

Western Blot Analysis:
1:500 dilution of this antibody detected BDNF on 10 µg of mouse brain lysates.

Target description

18 kDa, with a homodimer at ~27-30 kDa

Physical form

Rabbit polyclonal antiserum in buffer containing no preservatives. Lyophilized. Reconstitute with 50 μL of sterile distilled water.
Unpurified

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodrigo A P Martins et al.
Journal of neurochemistry, 95(1), 244-253 (2005-09-27)
Glutamate is a classical excitotoxin of the central nervous system (CNS), but extensive work demonstrates neuroprotective roles of this neurotransmitter in developing CNS. Mechanisms of glutamate-mediated neuroprotection are still under scrutiny. In this study, we investigated mediators of glutamate-induced neuroprotection
Chun-Lin Mai et al.
Molecular pain, 15, 1744806919826789-1744806919826789 (2019-01-12)
Accumulating evidence shows that inhibition of glycogen synthase kinase-3beta (GSK-3β) ameliorates cognitive impairments caused by a diverse array of diseases. Our previous work showed that spared nerve injury (SNI) that induces neuropathic pain causes short-term memory deficits. Here, we reported
Hao Zhang et al.
CNS neuroscience & therapeutics, 19(7), 484-493 (2013-03-29)
The intravenous anesthetic propofol caused episodic memory impairments in human. We hypothesized propofol caused episodic-like spatial memory retention but not acquisition impairments in rats and rescuing cAMP response element-binding protein (CREB) signaling using selective type IV phosphodiesterase (PDEIV) inhibitor rolipram
André Luis Bombeiro et al.
The European journal of neuroscience, 48(5), 2152-2164 (2018-08-14)
Peripheral nerve injuries severely impair patients' quality of life as full recovery is seldom achieved. Upon axonal disruption, the distal nerve stump undergoes fragmentation, and myelin breaks down; the subsequent regeneration progression is dependent on cell debris removal. In addition
Bohui Shu et al.
International journal of molecular medicine, 41(3), 1665-1672 (2017-12-30)
The pathogenesis of Alzheimer's disease (AD) has still not been fully elucidated, however it is thought that the build up of amyloid plaque at least partially causes the symptoms of AD. MicroRNAs (miRNAs) are endogenous non‑coding small RNA molecules that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service