Skip to Content
Merck
All Photos(1)

Key Documents

LP4

Sigma-Aldrich

Lipoprotein Deficient Serum, human, 100 mg

LPDS is isolated from fresh human serum by isopycnic ultracentrifugation, and is used for blocking of interfering antibodies.

Synonym(s):

LPDS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105901
eCl@ss:
26020100
NACRES:
NA.41

biological source

human

Quality Level

antibody form

serum

manufacturer/tradename

Chemicon®

technique(s)

blocking: suitable (interfering antibodies)

shipped in

dry ice

General description

LPDS is isolated from fresh human serum by isopycnic ultracentrifugation at the lower and upper density limits of the specified density range (Havel et al., 1955). Density adjustments are performed with solid KBr.

Concentration: 35 mg/mL

Source: Fresh human plasma. Tested negative for HBsAg and antibodies to HIV-I, HIV-II and Hepatitis C.

Quality

QUALITY ASSURANCE:

Protein determined by the Pierce BCA method with BSA as the standard. Purity of each preparation is verified by agarose gel electrophoresis using Fat Red 7B staining for lipid (2,3).

Physical form

Dulbecco′s PBS, pH 7.0, without Ca 2+. Filtered through a 0.22 micron filter. Not guaranteed sterile.

Storage and Stability

LPDS may be stored frozen and is quite stable. Cell culture medium containing 3-5 mg/mL of LPDS is approximately equivalent to medium containing 10% Fetal Bovine Serum.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M B Heaton et al.
The Journal of comparative neurology, 364(1), 113-120 (1996-01-01)
This study investigated the influence of ethanol exposure throughout gestation on cholinergic development within the rat striatal region. Pregnant Long-Evans rats were maintained on three diets throughout gestation: A liquid diet in which ethanol accounted for 35-39% of the total
Glucagon-like peptide-1 mobilizes intracellular Ca2+ and stimulates mitochondrial ATP synthesis in pancreatic MIN6 beta-cells.
Takashi Tsuboi,Gabriela da Silva Xavier,George G Holz,Laurence S Jouaville et al.
The Biochemical Journal null
Dominique Godfrin et al.
Journal of immunological methods, 326(1-2), 83-92 (2007-08-21)
A mixture of thiourea, urea and CHAPS (TUC) is an excellent solvent compatible with isoelectrofocusing (IEF) separation of water-insoluble protein extracts, and their subsequent two-dimensional gel electrophoresis is an important step in proteomic studies. The main aim of this work
Metabolic adaptation to protein restriction in insulin-dependent diabetes mellitus.
L J Hoffer,A Taveroff,A Schiffrin
The American Journal of Physiology null
Identification of a nuclear antigen with molecular weight of 48,000 differentially expressed in tumour and normal cells.
W M Krajewska,A Lipi?ska,M Marsza?ek,Z Kilia?ska,Z Wojtkowiak,L K?yszejko-Stefanowicz
Cell Biochemistry and Function null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service