Skip to Content
Merck
All Photos(1)

Key Documents

EGLP-35K

Millipore

Multi Species GLP-1 ELISA Kit

measures and quantifies active GLP-1 (7-36 amide and 7-37amide) in 100 μL plasma, serum or cell culture supernatent

Synonym(s):

GLP-1 (Active), Glucagon Like Peptide-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Product Name

Glucagon Like Peptide-1 (Active) ELISA, This Glucagon Like Peptide-1 (Active) ELISA is used to measure & quantify Glucagon Like Peptide-1 levels in Metabolism & Endocrine research.

Quality Level

species reactivity

mouse, rat, human

packaging

kit of 1 × 96 wells

parameter

100 μL sample volume (4hr assay)

assay range

accuracy: 86.9%
standard curve range: 2-100 pM

technique(s)

ELISA: suitable

input

sample type cell culture supernatant
sample type serum
sample type plasma (K2 EDTA)

NCBI accession no.

UniProt accession no.

application(s)

research use

detection method

fluorometric (355nm/460nm)

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GCG(2641)

General description

GLP-1 is secreted from gut endocrine cells in response to nutrient ingestion and plays multiple roles in metabolic homeostasis.

Specificity

Cross Reactivty
GLP-1 is conserved in almost all mammals. This kit may be used to mearure in such species (e.g. rat, mouse, primates, etc.)

Application

Overnight incubation at 4°C, followed by 2 hours 30 minutes at room temperature Sample volume 100 μl serum, plasma or tissue culture medium
Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders
Used to detect/quantify: Glucagon Like Peptide-1

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of oral fat perception by modified sham feeding on energy expenditure, hormones and appetite profile in the postprandial state.
Astrid J Smeets,Manuela P Lejeune,Margriet S Westerterp-Plantenga
The British Journal of Nutrition null
Rectal taurocholate increases L cell and insulin secretion, and decreases blood glucose and food intake in obese type 2 diabetic volunteers.
T E Adrian,S Gariballa,K A Parekh,S A Thomas,H Saadi,J Al Kaabi,N Nagelkerke,B Gedulin,A A Young
Diabetologia null
Activity-induced GLP-1 release in lean and obese subjects.
Tanja C M Adam, Margriet S Westerterp-Plantenga
Physiology & Behavior null
Compensation for obesity-induced insulin resistance in dogs: assessment of the effects of leptin, adiponectin, and glucagon-like peptide-1 using path analysis.
Verkest KR, Fleeman LM, Morton JM, Ishioka K, Rand JS
Domestic Animal Endocrinology null
GLP-1 Receptor Antagonist Exendin-(9-39) Elevates Fasting Blood Glucose Levels in Congenital Hyperinsulinism Owing to Inactivating Mutations in the ATP-Sensitive K+ Channel.
Andrew C Calabria,Changhong Li,Paul R Gallagher,Charles A Stanley,Diva D De Le
Diabetes null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service