Skip to Content
Merck
All Photos(3)

Key Documents

ECM550

Sigma-Aldrich

QCM ECMatrix Cell Invasion Assay, 24-well (8 µm), colorimetric

The CHEMICON Cell Invasion Assay Kit uses a 24-well plate, with 8 um pores, which provides an efficient system for evaluating the invasion of tumor cells through a basement membrane model.

Synonym(s):

Basement membrane cell invasion assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

vertebrates

manufacturer/tradename

Chemicon®
QCM

technique(s)

cell based assay: suitable

detection method

colorimetric

shipped in

wet ice

General description

Also available: Cell Comb Scratch Assay! Get biochemical data from a scratch assay!Click Here

Introduction
Invasion through the extracellular matrix (ECM) is an important step in tumor metastasis. Cancer cells initiate invasion by adhering to and spreading along the blood vessel wall. Proteolytic enzymes, such as MMP collagenases, dissolve tiny holes in the sheath-like covering (basement membrane) surrounding the blood vessels to allow cancer cells to invade.

The CHEMICON Cell Invasion Assay Kit provides an efficient system for evaluating the invasion of tumor cells through a basement membrane model. The kit utilizes ECMatrix, a reconstituted basement membrane matrix of proteins derived from the Engelbreth Holm-Swarm (EHS) mouse tumor. We examined the kit′s performance using human fibrosarcoma (HT-1080) and non-invasive fibroblasts (NIH3T3).

The CHEMICON Cell Invasion Assay Kit is ideal for evaluation of invasive tumor cells. Each CHEMICON Cell Invasion Assay Kit contains sufficient reagents for the evaluation of 12 samples.

The CHEMICON Cell Invasion Assay Kit is intended for research use only; not for diagnostic or therapeutic applications.

Test Principle:

The CHEMICON Cell Invasion Assay is performed in an Invasion Chamber, a 24-well tissue culture plate with 12 cell culture inserts. The inserts contain an 8 μm pore size polycarbonate membrane, over which a thin layer of ECMatrixTM is dried. The ECM layer occludes the membrane pores, blocking non-invasive cells from migrating through. Invasive cells, on the other hand, migrate through the ECM layer and cling to the bottom of the polycarbonate membrane.

Application:

The CHEMICON Cell Invasion Assay Kit is ideal for evaluation of invasive tumor cells. Each CHEMICON Cell Invasion Assay Kit contains sufficient reagents for the evaluation of 12 samples.

The CHEMICON Cell Invasion Assay Kit is intended for research use only; not for diagnostic or therapeutic applications.

Application

Detection of migrating cells is via crystal violet stain.
Research Category
Cell Structure

Packaging

12 wells

Components

Invasion Chamber: (Part No. 70019) One 24-well plate with 12 ECMatrixTM layered cell culture inserts.

Cell Stain: (Part No. 20294) One 10 mL bottle.

Cotton Swabs: (Part No. 10202) 24 each.

Forceps: (Part No. 10203) One each.

Storage and Stability

Store kit materials at 2-8°C for up to 6 months. Do not freeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10-13 - German Storage Class 10 to 13

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of a novel frizzled 8-related antiproliferative factor on in vitro carcinoma and melanoma cell proliferation and invasion.
Kristopher R Koch,Chen-Ou Zhang,Piotr Kaczmarek,Joseph Barchi,Li Guo,Hanief M Shahjee,Susan Keay
Investigational New Drugs null
RhoBTB2 (DBC2) functions as tumor suppressor via inhibiting proliferation, preventing colony formation and inducing apoptosis in breast cancer cells.
Mao H, Zhang L, Yang Y, Sun J, Deng B, Feng J, Shao Q, Feng A, Song B, Qu X
Gene null
Li Fan et al.
Experimental physiology, 105(5), 793-808 (2020-02-23)
What is the central question of this study? What is the role of lncRNA PRRT3-AS1 in the regulation of peroxisome proliferator-activated receptor γ (PPARγ) gene-mediated mechanistic target of rapamycin (mTOR) signalling pathway in proliferation, apoptosis and autophagy of prostate cancer
Down-regulation of JAK1 by RNA interference inhibits growth of the lung cancer cell line A549 and interferes with the PI3K/mTOR pathway.
Dan Liu,Yi Huang,Jing Zeng,Bojiang Chen,Na Huang,Na Guo,Lunxu Liu,Hong Xu,Xianming Mo,Weimin Li
Journal of Cancer Research and Clinical Oncology null
BCAR1 protein plays important roles in carcinogenesis and predicts poor prognosis in non-small-cell lung cancer.
Wei Huang,Bo Deng,Ru-Wen Wang,Qun-You Tan,Yong He,Yao-Guang Jiang,Jing-Hai Zhou
Testing null

Related Content

Discover cancer research resources with modeling and profiling tools for cell culture, genomics, biomarkers, and more to help maximize your cancer research.

Discover cancer research resources with modeling and profiling tools for cell culture, genomics, biomarkers, and more to help maximize your cancer research.

Discover cancer research resources with modeling and profiling tools for cell culture, genomics, biomarkers, and more to help maximize your cancer research.

Discover cancer research resources with modeling and profiling tools for cell culture, genomics, biomarkers, and more to help maximize your cancer research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service