Skip to Content
Merck
All Photos(1)

Documents

ABE387

Sigma-Aldrich

Anti-ATF4 Antibody

serum, from rabbit

Synonym(s):

Cyclic AMP-dependent transcription factor ATF-4, cAMP-dependent transcription factor ATF-4, Activating transcription factor 4, Cyclic AMP-responsive element-binding protein 2, CREB-2, cAMP-responsive element-binding protein 2

DNA-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATF4(468)

General description

Cyclic AMP-dependent transcription factor ATF-4 (ATF4) is a basic leucine-zipper (bZip) transcription factor, which regulates amino acid metabolism, DNA damage repair, chromatin remodeling, and apoptosis in response to cellular and ER stress. ATF4 works with various proteins, such as C/EBP homology protein (CHOP), aspargine synthetase (ASNS), and cAMP response element (CRE) among others to mediate cellular stress. ATF4 also regulates glucose homeostasis by suppressing beta-cell proliferation and insulin production. Furthermore, ATF4 targets the histone demethylase JMJD3 to alter chromatin structure and enhance gene transcription in response to amino acid deprivation.

Immunogen

Recombinant protein corresponding to human ATF4.

Application

Anti-ATF4 is an antibody targeting the ATF4 protein, validated for use in WB & ChIP.
Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory detected ATF4 in HEPG2 cell lysates (Su, N. and Kilberg, M. S., et al. (2008). J Biol Chem. 283(50):351016-35117.; Fu, L., et al. (2011). J Biol Chem. 286(42):36724-36738.).

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory detected ATF4 in HEPG2 and MEF cell lysates (Shan, J., et al. (2012). J Biol Chem. 287(43):36393-36403.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blot in untreated and HisOH treated HepG2 cell lysate.

Western Blot Analysis: A 1:2,000 dilution from a representative lot detected ATF4 in 10 µg of HisOH treated HepG2 cell lysate (HisOH was used to induce cellular stress mediated eIF2a phosphorylation, allowing for subsequent detection of ATF4. For enhanced performance, end-users may try higher dilutions of this antibody. For better results, 1% SDS was used during the secondary antibody incubation step.)

Target description

~48 kDa observed. The calculated molecular weight is 39 kDa; however, this protein may be observed at ~48 kDa in human tissue lysates and ~45 kDa in mouse cell lysates due to post-translational modification. Uncharacterized bands may be observed at ~57 kDa, ~59 kDa, ~75 kDa, and ~100 kDa in some cell lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HisOH treated HepG2 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dusana Majera et al.
The Prostate, 79(4), 352-362 (2018-12-01)
Castration-resistant prostate cancer (PCa) represents a serious health challenge. Based on mechanistically-supported rationale we explored new therapeutic options based on clinically available drugs with anticancer effects, including inhibitors of PARP1 enzyme (PARPi), and histone deacetylases (vorinostat), respectively, and disulfiram (DSF
Stefania Faletti et al.
Science translational medicine, 13(623), eabf7036-eabf7036 (2021-12-09)
Glioblastoma (GBM) is a fatal tumor whose aggressiveness, heterogeneity, poor blood-brain barrier penetration, and resistance to therapy highlight the need for new targets and clinical treatments. A step toward clinical translation includes the eradication of GBM tumor-initiating cells (TICs), responsible
Guochun Jiang et al.
mBio, 8(3) (2017-05-04)
Antiviral innate host defenses against acute viral infections include suppression of host protein synthesis to restrict viral protein production. Less is known about mechanisms by which viral pathogens subvert host antiviral innate responses for establishing their replication and dissemination. We
Katarina Chroma et al.
Cell death & disease, 13(3), 203-203 (2022-03-06)
Despite several approved therapeutic modalities, multiple myeloma (MM) remains an incurable blood malignancy and only a small fraction of patients achieves prolonged disease control. The common anti-MM treatment targets proteasome with specific inhibitors (PI). The resulting interference with protein degradation
Jungwoo Wren Kim et al.
eNeuro, 8(6) (2021-11-12)
The G2019S mutation in leucine-rich repeat kinase 2 (LRRK2) causes familial Parkinson's disease (PD) and is also found in a subset of idiopathic cases. Prior studies in Drosophila and human induced pluripotent stem cell (iPSC)-derived dopamine neurons uncovered a pronounced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service