Skip to Content
Merck
All Photos(2)

Key Documents

AB1560

Sigma-Aldrich

Anti-Opioid Receptor Antibody, δ, pain, NT

serum, Chemicon®

Synonym(s):

Anti-DOP, Anti-DOR, Anti-DOR1, Anti-OPRD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Recognizes Delta opioid receptor. The control peptide (catalog number AG288) is available for use in antibody blocking studies.

Immunogen

Delta opioid receptor N-terminal peptide (LVPSARAELQSSPLV).
Epitope: N-terminus

Application

Anti-Opioid Receptor Antibody, δ, pain, N-terminus detects level of Opioid Receptor & has been published & validated for use in FC, IH(P) & WB.
Immunohistochemistry on CNS tissues fixed in PLP: 1:1,000.

Reactive on paraffin embedded tissue.

Western Blot: 1:1000. Under reducing conditions the antibody will detect major bands at 36 and 72 kDa.

FACS on human T-cells (Sharp, 2001)

Optimal working dilutions must be determined by the end user.

IMMUNOHISTOCHEMISTRY PROTOCOL FOR AB1560

This antibody has been used successfully on 30 mm, free floating, 4% paraformaldehyde fixed rat brain tissue. All steps are performed under constant agitation. Suggested protocol follows.

1) 3 x 10 minute washes in TBS (without Triton).

2) Incubate for 30 minutes in TBS with 3% serum (same as host from secondary antibody).

3) Incubate primary antibody diluted appropriately in TBS with 1% serum (same as host from secondary antibody) (without Triton) for 2 hours at room temperature followed by 16 hours at 4°C.

4) 3 x 10 minute washes in TBS.

5) Incubate with secondary antibody diluted appropriately in TBS with1% serum (same as host from secondary antibody).

6) 3 x 10 minute washes in TBS.

7) ABC Elite (1:200 Vector Labs) in TBS.

8) 2 x 10 minute washes in TBS.

9) 1 x 10 minute wash in phosphate buffer (no saline).

10) DAB reaction with 0.06% NiCl added for intensification.

11) 2 x 10 minute washes in PBS.

12) 1 x 10 minute wash in phosphate buffer (no saline).
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Target description

36 and 72 kDa

Linkage

Replaces: MAB5330

Physical form

UnPurified whole rabbit antiSerum containing no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Spinal cord

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peripheral ?-opioid receptors attenuate the exercise pressor reflex.
Leal, AK; Yamauchi, K; Kim, J; Ruiz-Velasco, V; Kaufman, MP
American Journal of Physiology. Heart and Circulatory Physiology null
Yuan Xu et al.
Aging, 12(24), 25035-25059 (2020-11-17)
Our previous studies have shown that the δ-opioid receptor (DOR) is an important neuroprotector via the regulation of PTEN-induced kinase 1 (PINK1), a mitochondria-related molecule, under hypoxic and MPP+ insults. Since mitochondrial dysfunctions are observed in both hypoxia and MPP+
C M Cahill et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 21(19), 7598-7607 (2001-09-22)
Opioid receptors are known to undergo complex regulatory changes in response to ligand exposure. In the present study, we examined the effect of morphine on the in vitro and in vivo density and trafficking of delta opioid receptors (deltaORs). Prolonged
Sanoara Mazid et al.
Neurobiology of stress, 5, 37-53 (2016-12-17)
Drug addiction requires associative learning processes that critically involve hippocampal circuits, including the opioid system. We recently found that acute and chronic stress, important regulators of addictive processes, affect hippocampal opioid levels and mu opioid receptor trafficking in a sexually
Sarah M Gruba et al.
ACS measurement science Au, 2(1), 4-13 (2022-07-19)
Opioids are typically used for the treatment of pain related to disease or surgery. In the body, they enter the bloodstream and interact with a variety of immune and neurological cells that express the μ-, δ-, and κ-opioid receptors. One

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service