Skip to Content
Merck

SIC007

MISSION® siRNA Fluorescent Universal Negative Control #1, 6-FAM

Synonym(s):

MISSION® siRNA Fluorescent Universal Negative Control, 6-FAM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

color

Green (6-FAM)

storage temp.

−20°C

Application

Negative controls are an essential part of every siRNA experiment, and here we offer the siRNA Universal Negative Control #1 conjugated to 6-FAM for easy visualization and assessment of transfection efficiency.
Negative controls are an essential part of every siRNA experiment, and here we offer the siRNA Universal Negative Control #2 conjugated to Cyanine 3 for easy visualization and assessment of transfection efficiency.

Features and Benefits

  • 6-FAM labeled siRNA Universal Negative Control allows easy visualization of the siRNA in your cells by using fluorescence microscopy.
  • Rapidly assess transfection efficiency.
  • Ready to use.
  • Available in three sizes for your convenience.
  • Compatible with Human, Mouse, and Rat cells.

Legal Information

All siRNA oligos are sold under license from the Massachusetts Institute of Technology.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
SIC007-1NMOL04061826763384
SIC007-5X1NMOL04061826763391
SIC007-10NMOL04061826763377