Skip to Content
Merck
All Photos(1)

Key Documents

AB1032

Sigma-Aldrich

Anti-Versican Antibody, a.a. 535-598 of mouse versican

Chemicon®, from rabbit

Synonym(s):

GAG alpha Domain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... VCAN(1462)

General description

Versican is a large multi-domain chondroitin sulphate proteoglycan, is secreted by fibroblasts and other types of cells. It has a N-terminal hyaluronic acid (HA) binding domain. This domain has homology with the G1 and G2 domains of aggrecan, (G1 is the hyaluronic acid binding region), to cartilage link protein which stabilizes the binding of aggrecan to hyaluronic acid, and, to the hyaluronate receptor CD44. In addition, versican also has a lectin like domain, a C-terminal compliment repeat, and two EGF-like repeats, all of which can be found in aggrecan., Versican has an apparent molecular mass of ca. 1000 kDa, of which the core contributes 400 kDa from the 2409 amino acids, and the 12 -15 covalently attached chondroitin sulphate chains make up the rest.

Specificity

Mouse versican GAG alpha domain

Immunogen

Epitope: a.a. 535-598 of mouse versican
GST fusion protein containing amino acids 535-598 of mouse versican.

Application

Immunohistochemistry (formalin fixed, paraffin embedded): 10 μg/mL. Tissues must be treated with Chondroitinase before staining {0.5U/mL in 100mM Tris-HCL pH 7.2-7.4, 30 minutes at room temperature} Then block and stain.

Western blotting: 0.5 μg/mL

Immunoprecipitation

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Versican Antibody, a.a. 535-598 of mouse versican is validated for use in IP, WB, IH(P) for the detection of Versican.

Physical form

Format: Purified
Immunoglobulin was purified by affinity chromatography and is presented in PBS with no preservative Anti-GST has not been adsorbed.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Immunohistochemistry Positive control: surrounding cartilage, neural tube / brainWestern blotting positive control: newborn mouse brain

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina Hsiung et al.
Scientific reports, 8(1), 7530-7530 (2018-05-16)
The mechanisms responsible for macrovascular complications in diabetes remain to be fully understood. Recent studies have identified impaired vascular repair as a possible cause of plaque vulnerability in diabetes. This notion is supported by observations of a reduced content of
Expression of a novel versican variant in dorsal root ganglia from spared nerve injury rats.
Oliver Bogen et al.
Molecular pain, 15, 1744806919874557-1744806919874557 (2019-08-21)
Kelly L Short et al.
Journal of molecular endocrinology, 64(3), 155-164 (2020-01-21)
Glucocorticoid (GC) signaling via the glucocorticoid receptor (GR) is essential for lung maturation in mammals. Previous studies using global or conditional mouse model knockouts of the GR gene have established that GR-mediated signaling in the interstitial mesenchyme of the fetal
Maral Tajerian et al.
Molecular psychiatry, 23(12), 2302-2313 (2018-09-27)
Chronic pain poses a heavy burden for the individual and society, comprising personal suffering, comorbid psychiatric symptoms, cognitive decline, and disability. Treatment options are poor due in large part to pain centralization, where an initial injury can result in lasting
Simon J Foulcer et al.
Methods in molecular biology (Clifton, N.J.), 1229, 587-604 (2014-10-19)
Versican is a widely distributed chondroitin sulfate proteoglycan that forms large complexes with the glycosaminoglycan hyaluronan (HA). As a consequence of HA binding to its receptor CD44 and interactions of the versican C-terminal globular (G3) domain with a variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service