Skip to Content
Merck
All Photos(1)

Key Documents

8.20258

Sigma-Aldrich

5-Isopropyl-2-methylphenol

for synthesis

Synonym(s):

5-Isopropyl-2-methylphenol, 2-Methyl-5-isopropylphenol, Isothymol, Carvacrol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3)-5-[CH(CH3)2]C6H3OH
CAS Number:
Molecular Weight:
150.22
MDL number:
UNSPSC Code:
12352104
EC Index Number:
207-889-6
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

potency

810 mg/kg LD50, oral (Rat)

mp

3 °C

transition temp

flash point >100 °C

solubility

1.25 g/L

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Oc1c(ccc(c1)C(C)C)C

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4-7,11H,1-3H3

InChI key

RECUKUPTGUEGMW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.976 - 0.978
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service