Skip to Content
Merck
All Photos(1)

Key Documents

8.07051

Sigma-Aldrich

Propylene carbonate

for synthesis

Synonym(s):

Propylene carbonate, 4-Methyl-1,3-dioxolan-2-one, Propylene glycol carbonate, Carbonic acid propylene glycol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
MDL number:
UNSPSC Code:
12352115
EC Index Number:
203-572-1
NACRES:
NA.22
technique(s):
titration: suitable
bp:
243 °C/1013 hPa
vapor pressure:
0.04 hPa ( 20 °C)

vapor pressure

0.04 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

435 °C (DIN 51794)

potency

>5000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-14.3 % (v/v)

technique(s)

titration: suitable

pH

7.0 (20 °C, 200 g/L in H2O)

kinematic viscosity

1.4 cSt(43 °C)

bp

243 °C/1013 hPa

mp

-49 °C

transition temp

flash point 123 °C

solubility

240 g/L

density

1.20 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(COC1=O)C

InChI

1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3

InChI key

RUOJZAUFBMNUDX-UHFFFAOYSA-N

Related Categories

Application


  • Interfacial carbonyl groups of propylene carbonate facilitate the reversible binding of nitrogen dioxide.: Investigating the interaction of interfacial carbonyl groups in propylene carbonate with nitrogen dioxide, this study provides insights into the reversible binding mechanisms, important for developing better gas capture and storage systems (Clark & Allen, 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.204 - 1.206
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service