Skip to Content
Merck
All Photos(2)

Key Documents

752002

Sigma-Aldrich

Ethyl methyl carbonate

98%

Synonym(s):

Carbonic acid ethyl methyl ester, EMC, Methyl ethyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.378

density

1.006 g/mL at 25 °C

functional group

carbonate

SMILES string

CCOC(=O)OC

InChI

1S/C4H8O3/c1-3-7-4(5)6-2/h3H2,1-2H3

InChI key

JBTWLSYIZRCDFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl methyl carbonate (EMC) is the simplest asymmetric ester. It can be prepared via transesterification between dimethyl carbonate (DMC) and ethanol with mild reaction conditions. It is used as an intermediate for the synthesis of various compounds. Additionally, EMC is also used as an excellent co-solvent in the electrolyte of lithium-ion batteries.

Application

Ethyl methyl carbonate can be used as:      
  • A solvent for various reactions including esterification and amidation.   
  • An electrolyte in the development of graphite-based dual-ion batteries due to its ability reduces charge-transfer resistance, polarization, and self-discharge of graphite electrodes. It also enhances Coulombic efficiency, cycle, and rate performances.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.0 °F

Flash Point(C)

23.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu J, et al.
Electrochemical Communications, 13(3), 269-269 (2011)
McEwen A, et al.
Journal of the Electrochemical Society, 146(5), 1687-1687 (1999)
Susanne M Köhler et al.
Vaccine, 33(24), 2771-2777 (2015-04-29)
While proving highly effective in controlling Anthrax in farm animals all over the world currently attenuated live anthrax vaccines employed in a veterinary context suffer from drawbacks such as residual virulence, short term protection, variation in quality and, most importantly
McEwen A, et al.
Journal of the Electrochemical Society, 144(4), L84-L84 (1997)
Shamsudeen F Fagbo et al.
Emerging infectious diseases, 21(11), 1981-1988 (2015-10-21)
We investigated an outbreak of Middle East respiratory syndrome (MERS) at King Fahad Medical City (KFMC), Riyadh, Saudi Arabia, during March 29-May 21, 2014. This outbreak involved 45 patients: 8 infected outside KFMC, 13 long-term patients at KFMC, 23 health

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service