70978
BetaRed β-Galactosidase Assay Kit
Optimized reagents for ultrasensitive β-gal assays
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106513
NACRES:
NA.54
Recommended Products
General description
Optimized reagents for ultrasensitive β-gal assays
The BetaRed β-Gal Assay Kit is designed for rapid, sensitive measurement of β-galactosidase activity in cell extracts. The kit features Reportasol Extraction Buffer, which enables efficient passive extraction of cell protein while maintaining optimal enzyme activity, and is configured for 96-well format. The colorimetric BetaRed Assay is over 10-fold more sensitive (1 pg) than traditional ONPG-based assays and can be performed in conjunction with luciferase assays.
Components
•125 mgBetaRed Substrate
•25 mlReportasol Extraction Buffer
•100 mlReaction Buffer
•50 mlStop Buffer
•1.6 ml1 M DTT
•25 mlReportasol Extraction Buffer
•100 mlReaction Buffer
•50 mlStop Buffer
•1.6 ml1 M DTT
Warning
Toxicity: Multiple Toxicity Values, refer to MSDS (O)
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
YeastBuster™ Protein Extraction Reagent for Fast, Efficient Extraction of Proteins from Yeast
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service