Skip to Content
Merck
All Photos(1)

Key Documents

654270

Sigma-Aldrich

TNP-470

A cell-permeable Fumagillin analog that inhibits MetAP2, but not MetAP1, methionine aminopeptidase activity, without affecting the inhibitory activity of MetAP2 against HRI- catalyzed eIF-2α phosphorylation.

Synonym(s):

TNP-470, AGM-1470, O-(Chloroacetyl-carbamoyl)fumagillol, AGM1470, MetAP-2 Inhibitor II, MetAP2 Inhibitor II, Methionine Aminopeptidase 2 Inhibitor II, TNP470

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28ClNO6
CAS Number:
Molecular Weight:
401.88
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to light yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

ClCC(=O)NC(=O)O[C@H]1[C@H]([C@@H]([C@]3(OC3)CC1)[C@]2(O[C@@H]2CC=C(C)C)C)OC

InChI

1S/C19H28ClNO6/c1-11(2)5-6-13-18(3,27-13)16-15(24-4)12(7-8-19(16)10-25-19)26-17(23)21-14(22)9-20/h5,12-13,15-16H,6-10H2,1-4H3,(H,21,22,23)/t12-,13-,15-,16-,18+,19+/m1/s1

InChI key

MSHZHSPISPJWHW-PVDLLORBSA-N

General description

A cell-permeable Fumagillin (Cat. No. 344845) analog that inhibits MetAP2, but not MetAP1, methionine aminopeptidase activity (IC50 = 1 nM against human MetAP2) by covalently modifying MetAP2 active site histidine (aa 231 in human/mouse/rat; aa 174 in yeast) via its epoxide group, without affecting the inhibitory activity of MetAP2 against HRI- (heme-regulated inhibitor kinase) catalyzed eIF-2α phosphorylation. Cellular MetAP2 inhibition is particularly effective in inducing endothelial cytostasis (IC50 = 25 pM and 37 pM, respectively, against HUVEC and bovine BAEC proliferation by TNP-470), making TNP-470 a potent anti-angiogenesis agent in vivo (2 to 100 mg/kg in mice and rats; s.c.). Reported to be more potent and less toxic than Fumagillin.
A cell-permeable Fumagillin (Cat. No. 344845) analog that inhibits MetAP2, but not MetAP1, methionine aminopeptidase activity (IC50 = 1 nM against human MetAP2), without affecting the inhibitory activity of MetAP2 against HRI- (heme-regulated inhibitor kinase) catalyzed eIF-2α phosphorylation. A potent endothelial cytostasis inducer (IC50 = 25 pM and 37 pM, respectively, against HUVEC and bovine BAEC) in vitro and an effective anti-angiogenesis agent in vivo (2 to 100 mg/kg in mice and rats; s.c.).

Warning

Toxicity: Irritant (B)

Other Notes

Zhang, Y., et al. 2000. Proc. Natl. Acad. Sci. USA97, 6427.
Sin, N., et al. Proc. Natl. Acad. Sci. USA94, 6099.
Yamaoka, M., et al. 1993. Cancer Res.53, 4262.
Kusaka, M., et al. 1991. Biochem. Biophys. Res. Commun.174, 1070.
Ingber, D., et al. 1990. Nature348, 555.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service