Skip to Content
Merck
All Photos(1)

Key Documents

648469-M

Millipore

Troglitazone

A α-tocopherol (vitamin E) moiety containing thiazolidinedione class of insulin-sensitizer that acts as an activator of peroxisome proliferator-activated receptors γ (PPARγ).

Synonym(s):

Troglitazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO5S
CAS Number:
Molecular Weight:
441.54
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

description

RTECS - XJ5813130

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to yellow

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

storage temp.

−20°C

InChI

1S/C24H27NO5S/c1-13-14(2)21-18(15(3)20(13)26)9-10-24(4,30-21)12-29-17-7-5-16(6-8-17)11-19-22(27)25-23(28)31-19/h5-8,19,26H,9-12H2,1-4H3,(H,25,27,28)

InChI key

GXPHKUHSUJUWKP-UHFFFAOYSA-N

General description

A α-tocopherol (vitamin E) moiety containing thiazolidinedione class of insulin-sensitizer that acts as an activator of peroxisome proliferator-activated receptors γ (PPARγ). Also exhibits anti-proliferative and anti-inflammatory properties. Shown to induce cell cycle arrest and apoptosis in human hepatoma cells. Inhibits vascular smooth muscle cell (VSMC) migration by rapidly activating MEK/ERK pathway leading to the induction of c-fos and Ets-1 expression. Blocks PDGF-induced cell proliferation of pancreatic stellate and accelerates p21Cip1 turnover by inhibiting PKCδ signaling in VSMCs. Exerts positive inotropic effect via Ca2+ sensitization in the sarcoplasmic reticulum and inhibits Na+/H+ exchange activity and proliferation of macrovascular endothelial cells.

Biochem/physiol Actions

Primary Target
Peroxisome proliferator-activated receptors γ (PPARγ)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 6 months at -20°C. Ethanol stock solutions are stable for up to 3 months at -20°C.

Other Notes

Masamune, A., et al. 2002. J. Biol. Chem.277, 141.
Boyault, S., et al. 2001. FEBS Lett.501, 24.
de Dios, S.T., et al. 2001. J. Diabetes Complications15, 120.
Furuse, Y., et al. 2001. Br. J. Pharmacol.133, 1307.
Ghanim, H., et al. 2001. J. Clin. Endocrinol. Metab.86, 1306.
Goetze, S., et al. 2001. J. Cardiovasc. Pharmacol.38, 909.
Koga, H., et al. 2001. Hepatology33, 1087.
Rosen, E.D. and Spiegelman, B.M., 2001. J. Biol. Chem.276, 37731.
Subbaramaiah, K., et al. 2001. J. Biol. Chem.276, 12440.
Takashima, T., et al. 2001. Int. J. Oncol.19, 465.
Takeda, K., et al. 2001. J. Biol. Chem.276, 48950.
Tanaka, T., et al. 2001. Cancer Res.61, 2424.
Wakino, S., et al. 2001. J. Biol. Chem.276, 47650.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service