Skip to Content
Merck

565950

Sialyl Lewis x, Sodium Salt

Enzymatically prepared tetrasaccharide which acts as a minimal binding ligand for E-selectins.

Synonym(s):

Sialyl Lewis x, Sodium Salt, SLe x, Neu5Acα2,3Galβ1,4(Fucα1,3)GlcNAc, SLex, Neu5Acα2,3Galβ1,4(Fucα1,3)GlcNAc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H51N2NaO23
CAS Number:
Molecular Weight:
842.73
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 2 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N([C@H]1[C@@H](O[C@@](C[C@@H]1O)(O[C@@H]2[C@H]([C@@H](O[C@@H]([C@@H]2O)CO)O[C@@H]([C@H](O[C@@H]3O[C@H]([C@H]([C@H]([C@@H]3O)O)O)C)[C@@H](NC(=O)C)C=O)[C@H](O)CO)O)C(=O)O)[C@H](O)[C@H](O)CO)C(=O)C

InChI

1S/C31H52N2O23/c1-9-18(43)21(46)22(47)28(51-9)53-24(12(5-34)32-10(2)38)25(15(42)7-36)54-29-23(48)27(20(45)16(8-37)52-29)56-31(30(49)50)4-13(40)17(33-11(3)39)26(55-31)19(44)14(41)6-35/h5,9,12-29,35-37,40-48H,4,6-8H2,1-3H3,(H,32,38)(H,33,39)(H,49,50)/t9-,12-,13-,14+,15+,16+,17+,18+,19+,20-,21+,22-,23+,24+,25+,26+,27-,28-,29-,31-/m0/s1

InChI key

LAQPKDLYOBZWBT-NYLDSJSYSA-N

General description

Enzymatically prepared tetrasaccharide which acts as a minimal binding ligand for E-selectins. SLex reduces injury following ischemia and reperfusion and immune complex-induced acute lung injury in animal models. In both of these models, SLex is presumed to act by blocking E-selectin-mediated neutrophil recruitment to sites of inflammation.

Other Notes

Seekamp, A., et al. 1994. Am. J. Pathol.144, 592.
Mulligan, M.S., et al. 1993. J. Exp. Med.178, 623.
Ichikawa, Y., et al. 1992. J. Am. Chem. Soc. 114, 9283.
Lasky, L.A. 1992. Science 258, 964.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library