Skip to Content

S1782

3′-Sialyl-Lewis-X tetrasaccharide

Synonym(s):

α-NeuNAc-(2→3)-β-D-Gal-(1→4)(α-L-Fuc-[1→3])-D-GlcNAc, 3′-SLeX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H52N2O23
CAS Number:
Molecular Weight:
820.74
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

storage temp.

−20°C

SMILES string

CC1OC(OC(C(NC(C)=O)C=O)C(OC2OC(CO)C(O)C(OC3(CC(O)C(NC(C)=O)C(O3)C(O)C(O)CO)C(O)=O)C2O)C(O)CO)C(O)C(O)C1O

InChI

1S/C31H52N2O23/c1-9-18(43)21(46)22(47)28(51-9)53-24(12(5-34)32-10(2)38)25(15(42)7-36)54-29-23(48)27(20(45)16(8-37)52-29)56-31(30(49)50)4-13(40)17(33-11(3)39)26(55-31)19(44)14(41)6-35/h5,9,12-29,35-37,40-48H,4,6-8H2,1-3H3,(H,32,38)(H,33,39)(H,49,50)

InChI key

LAQPKDLYOBZWBT-UHFFFAOYSA-N

Biochem/physiol Actions

Antigen which binds to endothelial adhesion molecule, E-selectin.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library