Skip to Content
Merck
All Photos(1)

Key Documents

5.06304

Sigma-Aldrich

TrkB Antagonist, ANA-12

Synonym(s):

TrkB Antagonist, ANA-12, N-(2-((2-oxoazepan-3-yl)carbamoyl)phenyl)benzo[b]thiophene-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21N3O3S
CAS Number:
Molecular Weight:
407.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

10 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

[s]1c2c(cc1C(=O)Nc3c(cccc3)C(=O)NC4CCCCNC4=O)cccc2

InChI

1S/C22H21N3O3S/c26-20(25-17-10-5-6-12-23-21(17)27)15-8-2-3-9-16(15)24-22(28)19-13-14-7-1-4-11-18(14)29-19/h1-4,7-9,11,13,17H,5-6,10,12H2,(H,23,27)(H,24,28)(H,25,26)

InChI key

TUSCYCAIGRVBMD-UHFFFAOYSA-N

General description

A cell-permeable thiophene carboxymide derived compound that acts as a direct and selective antagonist of TrkB receptor. Binds to the high affinity site on the extracellular domain of TrkB in a dose-dependent manner (Kd = 10 nM) exhibiting much reduced binding to the low affinity site (Kd =12 µM) indicating that it does not compete with BDNF for the same binding site. Does not affect TrkA or TrkC receptors. Shown to permeate the blood-brain barrier, however, its antagonistic effects are not uniform throughout the brain. Does not affect the survivability of neurons. Reduces anxiety and depression related behavior in mice.

Biochem/physiol Actions

Primary Target
TrkB

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cazorla, M., et al. 2011. J. Clin. Invest.121,1846.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service