Skip to Content
Merck

Skip To

B3795

Brain-derived neurotrophic factor human

≥95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Abrineurin, BDNF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Biological source:
human
Recombinant:
expressed in E. coli
Assay:
≥95% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
~27 kDa
Impurities:
≤1.00 EU/μg Endotoxin level
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Brain-derived neurotrophic factor human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

lyophilized powder

quality

endotoxin tested

mol wt

~27 kDa

packaging

pkg of 10 μg, pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

technique(s)

cell culture | mammalian: suitable

impurities

≤1.00 EU/μg Endotoxin level

UniProt accession no.

storage temp.

−20°C

General description

BDNF is a member of the neurotrophin family of growth factors that includes NGF, NT-3, and NT-4. All neurotrophins have six conserved cysteine residues and share a 55% sequence identity at the amino acid level. BDNF has been shown to enhance the survival and differentiation of several classes of neurons in vitro, including neural crest and placode-derived sensory neurons, dopaminergic neurons in the substantia nigra, basal forebrain cholinergic neurons, hippocampal neurons, and retinal ganglial cells.1 BDNF is expressed within peripheral ganglia and is not restricted to neuronal target fields, raising the possibility that BDNF has paracrine or even autocrine actions on neurons as well as non-neuronal cells.
The active form of BDNF recombinant human protein (27 kDa) is a dimer formed by two identical 119 amino acid subunits held together by strong hydrophobic interactions

Application

Brain-derived neurotrophic factor (BDNF) recombinant human protein has been used as a supplement in various types of culture media.[1][2][3]

Biochem/physiol Actions

Brain-derived neurotrophic factor (BDNF) is a 13.6 kDa (or 27.2 kDa dimer) member of the neurotrophin family. BDNF has identical amino acid sequence in human, mouse, and pig with full cross-reactivities. BDNF is important in development and maintenance of neuronal populations within the central nervous system or cells directly associated with it. BDNF has been shown to enhance the survival and differentiation of several classes of neurons in vitro, including neural crest and placode-derived sensory neurons, dopaminergic neurons in the substantia nigra, basal forebrain cholinergic neurons, hippocampal neurons, and retinal ganglial cells.

Physical form

lyophilized from a sterile 0.2 μm filtered solution with BSA

Analysis Note

The biological activity is determined by its ability to stimulate the proliferation of human neuroblastoma cells, SH-SY5Y.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRP3014G1777SRP6053
biological source

human

biological source

human

biological source

human

biological source

human

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥98% (SDS-PAGE)

assay

≥95% (HPLC), ≥95% (SDS-PAGE)

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

form

lyophilized powder

form

lyophilized

form

lyophilized powder

form

lyophilized


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters