Skip to Content
Merck
All Photos(1)

Key Documents

5.05601

Sigma-Aldrich

LRH-1 Antagonist

Synonym(s):

LRH-1 Antagonist, Liver Receptor Homologue 1 Antagonist, NR5A2 Antagonist, Nuclear Receptor Subfamily 5 Group A Member 2 Antagonist, 1-(3ʹ-(1-(2-Morpholinoethyl)-1H-pyrazol-3-yl)biphenyl-3-yl)ethanone, 1-(3ʹ-(1-(2-(4-Morpholinyl)ethyl)-1H-pyrazol-3-yl)-3-biphenylyl)ethanone, Nuclear Receptor Subfamily 5 Group A Member 2 Antagonist, 1-(3ʹ-(1-(2-Morpholinoethyl)-1H-pyrazol-3-yl)biphenyl-3-yl)ethanone, 1-(3ʹ-(1-(2-(4-Morpholinyl)ethyl)-1H-pyrazol-3-yl)-3-biphenylyl)ethanone, Liver Receptor Homologue 1 Antagonist, NR5A2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N3O2
CAS Number:
Molecular Weight:
375.46
MDL number:
UNSPSC Code:
41106200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyrazolylbiphenylethanone compound that targets LRH-1/NR5A2 ligand binding domain (LBD) via direct affinity interaction (Kd = 1.5 µM), preventing LRH-1 from assuming an active conformation. Shown to antagonize LRH-1-, but not SF-1-, dependent G0S2 transcription (IC50 = 5 µM vs. no inhibition at 10 µM, respectively; 24 h drug treatment commences 3 h after tetracycline induction in HEK293 transfectants), while exhibiting no inhibitory effect against AR (androgen hormone receptor), ERα (estrogen hormone receptor α), or TRβ (thyroid hormone receptor β) transcription activity in HeLa-based reporter transactivation assays at concentrations up to 10 µM. Reported to exhibit non-cytotoxic antiproliferation activity in LRH-1-expressing cancer cultures (Cell line/GI50 = AsPC-1/20 µM, HT-29/15 µM, MDA-MB-468/20 µM, T47D/20 µM), while being much less effective against the proliferation of pancreatic cancer L3.3 line that does not express LRH-1 (20% inhibition at 40 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LRH-1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Benod, C., et al. 2013. J. Biol. Chem.288, 19830.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service