Skip to Content
Merck
All Photos(1)

Key Documents

476485

Sigma-Aldrich

PKA Inhibitor 14-22 Amide, Cell-Permeable, Myristoylated

PKA Inhibitor 14-22 Amide is myristoylated at the N-terminus that enhances its cell-permeability. The non-myristoylated version is shown to be a specific inhibitor of PKA (Ki = 36 nM).

Synonym(s):

PKA Inhibitor 14-22 Amide, Cell-Permeable, Myristoylated, PKI 14-22 Amide, Cell-Permeable, Myristoylated Protein Kinase A Inhibitor Amide 14-22, Cell-Permeable, Myr-GRTGRRNAI-NH₂, Protein Kinase A Inhibitor 14-22 Amide, PKA Inhibitor XIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H100N20O12
Molecular Weight:
1209.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Heat-stable protein kinase inhibitor (PKI) peptide sequence (14-22) that has been myristoylated at the N-terminus, enhancing its cell-permeability. The non-myristoylated version of this peptide is a highly specific inhibitor (Ki = 36 nM) of cAMP-dependent protein kinase (PKA).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cAMP-dependent protein kinase
Product does not compete with ATP.
Reversible: no
Target Ki: 36 nMf or cAMP-dependent protein kinase

Warning

Toxicity: Standard Handling (A)

Sequence

Myr-N-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Paman, K., et al. 1998. J. Lipid Res. 39, 1091.
Rimon, G., and Rubin M. 1998. Biochim. Biophys. Acta 1380, 289.
Harris, T.E., et al. 1997. Biochem. Biophys. Res. Commun. 232, 648.
Muniz, M., et al. 1997. Proc. Natl. Acad. Sci. USA 94, 14461.
Zoukhri, D., et al. 1997. Am. J. Physiol. 272, C263.
Eichholtz, T., et al. 1993. J. Biol. Chem.268, 1982.
Ward, N.E. and O’Brian, C.A. 1993. Biochemistry32, 11903.
Walsh, D.A. and Glass, D.B. 1991. Methods Enzymol. 201, 304.
Glass, D.B., et al. 1989. J. Biol. Chem.264, 8802.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duomeng Yang et al.
Communications biology, 5(1), 96-96 (2022-01-27)
Intrinsic cardiac adrenergic (ICA) cells regulate both developing and adult cardiac physiological and pathological processes. However, the role of ICA cells in septic cardiomyopathy is unknown. Here we show that norepinephrine (NE) secretion from ICA cells is increased through activation
Lian-Pan Wu et al.
Acta pharmacologica Sinica, 41(1), 34-46 (2019-09-14)
Abnormal growth of the intimal layer of blood vessels (neointima formation) contributes to the progression of atherosclerosis and in-stent restenosis. Recent evidence shows that the 18-kDa translocator protein (TSPO), a mitochondrial membrane protein, is involved in diverse cardiovascular diseases. In
Lukasz Buldak et al.
Journal of applied biomedicine, 20(4), 130-140 (2023-01-29)
Statins are primary drugs in the treatment of hyperlipidemias. This group of drugs is known for its beneficial pleiotropic effects (e.g., reduction of inflammatory state). However, a growing body of evidence suggests its diabetogenic properties. The culpable mechanism is not
Akiko Nakayama et al.
JCI insight, 5(23) (2020-12-04)
Atherosclerosis develops preferentially in areas of the arterial system, in which blood flow is disturbed. Exposure of endothelial cells to disturbed flow has been shown to induce inflammatory signaling, including NF-κB activation, which leads to the expression of leukocyte adhesion
Irma E Cisneros et al.
Frontiers in neurology, 11, 593146-593146 (2020-12-17)
Methamphetamine (METH) use, referred to as methamphetamine use disorder (MUD), results in neurocognitive decline, a characteristic shared with HIV-associated neurocognitive disorders (HAND). MUD exacerbates HAND partly through glutamate dysregulation. Astrocyte excitatory amino acid transporter (EAAT)-2 is responsible for >90% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service