SCP0064
cAMP Dependent PKI Peptide
≥95% (HPLC), lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
product name
cAMP Dependent Protein Kinase Inhibitor PKI-tide,
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥67%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Ile-Ala-Ala-Gly-Arg-Thr-Gly-Arg-Arg-Gln-Ala-Ile-His-Asp-Ile-Leu-Val-Ala-Ala
Application
IAAGRTGRRQAIHDILVAA is used as a cAMP-dependent protein kinase inhibitory peptide.
Biochem/physiol Actions
cAMP dependent protein kinase inhibitors are generally designed to target the synthesis and degradation of cAMP (cyclic adenosine monophophaphate), a second messenger.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service