Skip to Content
Merck
All Photos(2)

Key Documents

341216

Sigma-Aldrich

NSC668394

≥95% (HPLC), solid, Ezrin inhibitor, Calbiochem®

Synonym(s):

Ezrin Inhibitor, NSC668394, 7-(3,5-dibromo-4-hydroxyphenethylamino)quinoline-5,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12Br2N2O3
CAS Number:
Molecular Weight:
452.10
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Ezrin Inhibitor, NSC668394, The Ezrin Inhibitor, NSC668394 controls the biological activity of Ezrin.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red-orange

solubility

DMSO: 50 mg/mL, clear, red-orange

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1C=C(NCCC2=CC(Br)=C(O)C(Br)=C2)C(C3=C1C=CC=N3)=O

InChI key

OHPHDPYJEHZGDE-UHFFFAOYSA-N

General description

A cell-permeable, quinoline that binds directly to ezrin (Kd = 12.6 µM) in a reversible manner, and inhibits PKCι-mediated ezrin phosphorylation at Thr567 (IC50 = 8.1 µM) and ezrin-actin binding, in vitro. It also inhibits murine osteosarcoma K7M2 cell invasion in a monolayer of HUVEC cells, dose-dependently, from 1 to 10 µM. At 10 µM, it causes a unique developmental defect in zebrafish, characterized by reduced motility of eye progenitor cells. Furthermore, this small molecule prevents metastatic growth in an ex vivo mouse lung organ culture assay at 10 µM and attenuates ezrin-dependent OS metastatic growth in mouse lung model in vivo at 2.26 mg/kg/day.
A cell-permeable, quinoline that binds directly to ezrin (Kd = 12.6 µM) in a reversible manner, and inhibits PKCι-mediated ezrin phosphorylation at Thr567 (IC50 = 8.1 µM) and ezrin-actin binding, in vitro. It also inhibits murine osteosarcoma K7M2 cell invasion in a monolayer of HUVEC cells, dose-dependently, from 1 to 10 µM. At 10 µM, it causes a unique developmental defect in zebrafish, characterized by reduced motility of eye progenitor cells. Furthermore, this small molecule prevents metastatic growth in an ex vivo mouse lung organ culture assay at 10 µM and attenuates ezrin-dependent OS metastatic growth in mouse lung model in vivo at 2.26 mg/kg/day.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bulut, G., et al. 2011. Oncogene31, 269.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yueli Tian et al.
Experimental physiology, 108(5), 740-751 (2022-09-27)
What is the central question of this study? To reveal the role and biological mechanism of PDPN in the progression of gastric cancer. What is the main finding and its importance? This study focused on a prognostic predictor, PDPN, which
Henry De Belly et al.
Cell, 186(14), 3049-3061 (2023-06-14)
Membrane tension is thought to be a long-range integrator of cell physiology. Membrane tension has been proposed to enable cell polarity during migration through front-back coordination and long-range protrusion competition. These roles necessitate effective tension transmission across the cell. However
Samuel J Ghilardi et al.
Molecular biology of the cell, 32(18), 1707-1723 (2021-07-01)
Interactions between the actin cytoskeleton and the plasma membrane are important in many eukaryotic cellular processes. During these processes, actin structures deform the cell membrane outward by applying forces parallel to the fiber's major axis (as in migration) or they
Agata Nyga et al.
iScience, 26(5), 106696-106696 (2023-05-12)
Animal cells undergo repeated shape changes, for example by rounding up and respreading as they divide. Cell rounding can be also observed in interphase cells, for example when cancer cells switch from a mesenchymal to an ameboid mode of cell
Chien-Ting Wu et al.
Cell, 186(1), 112-130 (2022-12-30)
How SARS-CoV-2 penetrates the airway barrier of mucus and periciliary mucins to infect nasal epithelium remains unclear. Using primary nasal epithelial organoid cultures, we found that the virus attaches to motile cilia via the ACE2 receptor. SARS-CoV-2 traverses the mucus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service