Skip to Content
Merck
All Photos(1)

Key Documents

182515

Sigma-Aldrich

CK-666

≥95% (HPLC), solid, Arp2/3 complex inhibitor, Calbiochem

Synonym(s):

Arp2/3 Complex Inhibitor I, CK-666, 2-Fluoro-N-(2-(2-Methyl-1H-indol-3-yl)ethyl)benzamide, CK-0944666, Actin Assembly Inhibitor XIII, Actin Assembly Inhibitor XIII, 2-Fluoro-N-(2-(2-Methyl-1H-indol-3-yl)ethyl)benzamide, CK-0944666

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17FN2O
CAS Number:
Molecular Weight:
296.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Arp2/3 Complex Inhibitor I, CK-666, Arp2/3 Complex Inhibitor I, CK-666, CAS 442633-00-3, is a cell-permeable selective inhibitor of actin assembly mediated by actin-related protein Arp2/3 complex (IC50 = 4 uM in human).

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(cccc1)C(=O)NCCc2c3c([nH]c2C)cccc3

InChI

1S/C18H17FN2O/c1-12-13(14-6-3-5-9-17(14)21-12)10-11-20-18(22)15-7-2-4-8-16(15)19/h2-9,21H,10-11H2,1H3,(H,20,22)

InChI key

UXRKUKRXVWJFER-UHFFFAOYSA-N

General description

A cell-permeable indolyl-fluorobenzamide compound that selectively inhibits actin assembly mediated by actin-related protein Arp2/3 complex of human, bovine, fission yeast S. pombe, and budding yeast S. cerevisiae origin (IC50 = 4, 17, 5, and 12 µM, respectively), without affecting S. pombe formin domain Cdc12(FH2)-mediated or the spontaneous actin polymerization. Shown to inhibit the actin filament "comet tails" formation around intracellular Listeria in infected SKOV3 cells (IC50 = 7 µM) in a reversible manner. Structrual studies indicate that CK-666 targets a pocket formed between subdomain 4 of Arp2 and subdomain 1 of Arp3, preventing Arp2/3 from shifting into an active conformation upon N-WASP-VCA binding. CK-666 and CK-869 (Cat. No. 182516) exhibit different modes of binding, resulting in their different yeast cross-reactivities. CK-689 (Cat. No. 182517) can serve as a negative control.
A cell-permeable indolyl-fluorobenzamide compound that selectively inhibits actin assembly mediated by actin-related protein Arp2/3 complex of human, bovine, fission yeast S. pombe, and budding yeast S. cerevisiae origin (IC50 = 4, 17, 5, and 12 µM, respectively), without affecting S. pombe formin domain Cdc12(FH2)-mediated or the spontaneous actin polymerization. Shown to inhibit the actin filament "comet tails" formation around intracellular Listeria in infected SKOV3 cells (IC50 = 7 µM) in a reversible manner. Structrual studies indicate that CK-666 targets a pocket formed between subdomain 4 of Arp2 and subdomain 1 of Arp3, preventing Arp2/3 from shifting into an active conformation upon N-WASP-VCA binding. CK-666 and CK-869 (Cat. No. 182516) exhibit different modes of binding, resulting in their different yeast cross-reactivities. CK-689 (Cat. No. 182517) can serve as a negative control.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Nolen, B.J., et al. 2009. Nature460, 1031.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Binh An Truong Quang et al.
Nature communications, 12(1), 6511-6511 (2021-11-13)
In animal cells, shape is mostly determined by the actomyosin cortex, a thin cytoskeletal network underlying the plasma membrane. Myosin motors generate tension in the cortex, and tension gradients result in cellular deformations. As such, many cell morphogenesis studies have
Kathleen Scheffler et al.
Journal of cell science, 135(6) (2022-02-04)
Formation of healthy mammalian eggs from oocytes requires specialised F-actin structures. F-actin disruption produces aneuploid eggs, which are a leading cause of human embryo deaths, genetic disorders and infertility. We found that oocytes contain prominent nuclear F-actin structures that are
Xarxa Quiroga et al.
eLife, 12 (2023-09-25)
As cells migrate and experience forces from their surroundings, they constantly undergo mechanical deformations which reshape their plasma membrane (PM). To maintain homeostasis, cells need to detect and restore such changes, not only in terms of overall PM area and
Eoin McEvoy et al.
Nature communications, 13(1), 7089-7089 (2022-11-20)
The formation and recovery of gaps in the vascular endothelium governs a wide range of physiological and pathological phenomena, from angiogenesis to tumor cell extravasation. However, the interplay between the mechanical and signaling processes that drive dynamic behavior in vascular
An ancestral mycobacterial effector promotes dissemination of infection.
Saelens, et al.
Cell, 185, 4507-4525 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service