Skip to Content
Merck
All Photos(2)

Key Documents

1.88007

Supelco

CombiSolvent Keto

suitable for Karl Fischer Titration, Aquastar®

Synonym(s):

Ketone Solvent Blend

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601
NACRES:
NA.21

Product Name

CombiSolvent Keto, suitable for Karl-Fischer titration

product line

Aquastar®

Quality Level

form

liquid

reaction suitability

reaction type: volumetric

technique(s)

Karl-Fischer titration: suitable
titration: suitable

transition temp

flash point 31 °C

density

0.924 g/cm3 at 20 °C

format

single component solution

storage temp.

15-25°C

General description

Solvent for volumetric Karl Fischer Titration with one component reagents for aldehydes and ketones Aquastar®

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

87.8 °F

Flash Point(C)

31 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Methods including internal extraction, external extraction, and Karl Fischer oven ensure accurate water content determination by titration.

Methods including internal extraction, external extraction, and Karl Fischer oven ensure accurate water content determination by titration.

Methods including internal extraction, external extraction, and Karl Fischer oven ensure accurate water content determination by titration.

Protocols

Summary application report for analysis of moisture in Acetone

Summary application report for analysis of moisture in Methyl ethyl ketone, 2-Butanone.

Summary application report for analysis of moisture in Formic acid

Summary application report for analysis of moisture in 1-Methyl-2-pyrrolidone.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service