1.88002
CombiTitrant 2
1 ml ≙ ca. 2 mg H₂O, one-component reagent for Karl Fischer titration, Aquastar™
Synonym(s):
CombiTitrant 2
About This Item
Recommended Products
Product Name
CombiTitrant 2,
product line
Aquastar®
Quality Level
form
liquid
shelf life
36 mo. (from manufacture date)
reaction suitability
reaction type: volumetric
concentration
2 mg/mL
technique(s)
Karl-Fischer titration: suitable
titration: suitable
color
red-brown
format
single component solution
shipped in
ambient
storage temp.
15-25°C
Application
1.88002.1045 is packaged in GL45 poly coated glass bottle
1.88002.2538 is packaged in 38/430 poly coated glass bottle
1.88002.2545 is packaged in GL45 poly coated glass bottle
Packaging
Analysis Note
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Oral
Target Organs
Thyroid
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Summary application report for analysis of moisture in 1-Methyl-2-pyrrolidone.
Summary application report for analysis of moisture in 1-Methyl-2-pyrrolidone.
Summary application report for analysis of moisture in Formic acid
Summary application report for analysis of moisture in 1-Methyl-2-pyrrolidone.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service