Skip to Content
Merck
All Photos(1)

Key Documents

1.46432

Millipore

Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules

tube capacity × filling volume 17 mL × 9 mL, closure type, blue screw cap with 2 loci, box of 100 tubes

Synonym(s):

CASO, Casein Soybean Digest Broth, SCDB, TSB, Tryptone Soya Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106204
NACRES:
NA.74

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type blue screw cap with 2 loci
ready-to-use

packaging

box of 100 tubes

capacity

17 mL

tube capacity × filling volume

17 mL × 9 mL

color

clear yellow Media

pH

7.3

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic soy broth is suitable for the isolation and cultivation of fastidious aerobic bacteria, yeasts, and molds. Two highly nutritious peptones support the growth of a wide variety of aerobic and facultative anaerobic microorganisms, including fungi.

Application

Tryptic Soy Broth is used for sterility testing of non-sterile substances, preparations, products, as well as a pre-enrichment broth to test the specified microorganisms or determine total viable aerobic count using the MPN method.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service