Skip to Content
Merck
All Photos(1)

Key Documents

T9032

Millipore

Thioglycollate medium

Microbiologically tested, NutriSelect® Plus

Synonym(s):

FTG, Fluid Thioglycolate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

Quality Level

sterility

non-sterile

form

powder

manufacturer/tradename

NutriSelect® Plus

application(s)

clinical testing
environmental
food and beverages
sterility testing

suitability

bacteria

Application

Powdered broth medium for the cultivation of aerobic and anaerobic microorganisms.

Preparation Note

Solve 29.8 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Borong Zhou et al.
Nature microbiology, 5(12), 1576-1587 (2020-10-21)
Sepsis is the most common cause of death for patients in intensive care worldwide due to a dysregulated host response to infection. Here, we investigate the role of sequestosome-1 (SQSTM1/p62), an autophagy receptor that functions as a regulator of innate
Yan Wang et al.
European journal of pharmacology, 890, 173630-173630 (2020-10-13)
Glucocorticoids are commonly used in clinic, but the immunosuppression seriously hinders their usage. Herein, immunomodulatory effect of artesunate (AS) on hydrocortisone (HC)-induced immunosuppression was investigated. HC-induced immunosuppression mice (HC mice) were established by intramuscular administration with HC (20 mg/kg) once a
Xiaojiao Wang et al.
Signal transduction and targeted therapy, 5(1), 35-35 (2020-04-17)
The inhibitory receptor signal regulatory protein-α (Sirpα) is a myeloid-specific immune checkpoint that engages the "don't eat me" signal CD47, which is expressed on tumor and normal tissue cells. However, the profile and regulatory mechanism of Sirpα expression in tumor-associated
Hongming Miao et al.
Nature communications, 7, 11716-11716 (2016-05-18)
Metabolic reprogramming in stromal cells plays an essential role in regulating tumour growth. The metabolic activities of tumour-associated macrophages (TAMs) in colorectal cancer (CRC) are incompletely characterized. Here, we identify TAM-derived factors and their roles in the development of CRC.
David Frommhold et al.
The Journal of experimental medicine, 205(6), 1435-1446 (2008-06-04)
Recent in vitro studies have suggested a role for sialylation in chemokine receptor binding to its ligand (Bannert, N., S. Craig, M. Farzan, D. Sogah, N.V. Santo, H. Choe, and J. Sodroski. 2001. J. Exp. Med. 194:1661-1673). This prompted us

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service