Skip to Content
Merck
All Photos(1)

Key Documents

1.07979

Supelco

Thiourea

GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Thiourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4N2S
CAS Number:
Molecular Weight:
76.12
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-543-5
NACRES:
NB.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

autoignition temp.

440 °C (Dust)

potency

1750 mg/kg LD50, oral (Rat)
>2800 mg/kg LD50, skin (Rabbit)

pH

6-8 (20 °C, 50 g/L in H2O)

mp

169-173 °C

solubility

137 g/L

density

1.405 g/cm3 at 20 °C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay: ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance of solution (50 g/l; water): Clear to almost clear and colorless.
Melting point: 174 - 178 °C
Fe (Iron): ≤ 0.0005 %
Sulfated ash (600 °C): ≤ 0.1 %
Loss on Drying (105°C): ≤ 0.5 %
Sensitivity to bismuth: passes test
Corresponds to ACS, Reag. Ph Eur

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Select a Product Number

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Customers Also Viewed

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service