1.06384
Sodium carbonate decahydrate
EMPROVE® ESSENTIAL, Ph. Eur., BP, E 500
Pharma Manufacturing
Synonym(s):
Sodium carbonate decahydrate, Soda, Sodium Carbonate decahydrate
About This Item
Recommended Products
grade
reagent
Quality Level
Agency
BP
Ph. Eur.
product line
EMPROVE® ESSENTIAL
form
solid
pH
11-12 (25 °C, 50 g/L in H2O)
mp
33 °C (Elimination of water of crystallisation)
solubility
210 g/L
density
1.44 g/cm3 at 20 °C
application(s)
liquid formulation
pharmaceutical
solid formulation
storage temp.
15-25°C
InChI
1S/CH2O3.2Na.10H2O/c2-1(3)4;;;;;;;;;;;;/h(H2,2,3,4);;;10*1H2/q;2*+1;;;;;;;;;;/p-2
InChI key
XYQRXRFVKUPBQN-UHFFFAOYSA-L
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service