1.03028
2,6-Dichlorophenol-indophenol sodium salt dihydrate
GR for analysis for the determination of ascorbic acid
Synonym(s):
2,6-Dichlorophenol-indophenol sodium salt dihydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H6Cl2NNaO2 · 2H2O
CAS Number:
Molecular Weight:
326.11
MDL number:
UNSPSC Code:
12352107
EC Index Number:
210-640-4
NACRES:
NA.21
Recommended Products
Quality Level
bulk density
320 kg/m3
storage temp.
15-25°C
SMILES string
[Na+].Clc1c(c(cc(c1)N=C2C=CC(=O)C=C2)Cl)[O-]
InChI
1S/C12H7Cl2NO2.Na/c13-10-5-8(6-11(14)12(10)17)15-7-1-3-9(16)4-2-7;/h1-6,17H;/q;+1/p-1
InChI key
FHLDWQLHDYCXKI-UHFFFAOYSA-M
Analysis Note
Assay (perchloric acid titration, calculated on dried substance): ≥ 98
Identity (IR-spectrum): passes test
TLC-Test: passes test
Foreign coloring agents: passes test
Loss on drying (120 °C): ≤ 12.0 %
Identity (IR-spectrum): passes test
TLC-Test: passes test
Foreign coloring agents: passes test
Loss on drying (120 °C): ≤ 12.0 %
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service