Skip to Content
Merck
All Photos(1)

Key Documents

1.01566

Millipore

L-Asparagine monohydrate

for biochemistry

Synonym(s):

L-Asparagine monohydrate, Asparaginic acid semiamide, Asn

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O3 · xH2O
CAS Number:
Molecular Weight:
132.12 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-735-9
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

form

solid

pH

4.0-5.5 (20 °C, 20 g/L in H2O)

mp

215-217 °C (decomposes)

solubility

22 g/L

bulk density

530 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3][C@@H](CC(=O)N)C(=O)[O-].O

InChI

1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m0./s1

InChI key

RBMGJIZCEWRQES-DKWTVANSSA-N

Application


  • Chiral Resolution and Amino Acid Racemase Applications: L-Asparagine monohydrate has been utilized in the immobilization of amino acid racemases complemented with preferential crystallization process for enhanced chiral resolutions (Carneiro et al., 2020).

  • Electro-optical Property Studies: L-Asparagine monohydrate can be used to prepare growth crystals to study its electro-optical properties, particularly in noncentrosymmetric crystal environments. (Shkir et al., 2015).


Analysis Note

  • Assay (perchloric acid titration, calculated on dried substance): 99.0 - 101.0 %
  • Identity (IR-spectrum): passes test
  • Identity (specific rotation): passes test
  • Appearance: white to almost white crystalline powder or colorless crystals
  • Appearance of solution (20 g/l, CO₂-free water): clear and colorless
  • Spec. rotation (α 20/D, 100 g/l, hydrochloric acid 3 mol/l, calc. on dried substance): +33.7 to +36.0
  • Chloride (Cl): ≤ 50 ppm
  • Sulfate (SO₄): ≤ 50 ppm
  • As (Arsenic): ≤ 5 ppm
  • Ca (Calcium): ≤ 10 ppm
  • Co (Cobalt): ≤ 5 ppm
  • Fe (Iron): ≤ 5 ppm
  • K (Potassium): ≤ 5 ppm
  • Mg (Magnesium): ≤ 5 ppm
  • Na (Sodium): ≤ 50 ppm
  • Pb (Lead): ≤ 10 ppm
  • Zn (Zinc): ≤ 5 ppm
  • Ninhydrin-positive substances (LC)(Aspartic acid): ≤ 0.5 %
  • Ninhydrin-positive substances (LC)(any other ninhydrin-positive impurity): ≤ 0.3 %
  • Ninhydrin-positive substances (LC) (ammonium (570 nm)): ≤ 0.02 %
  • Ninhydrin-positive substances (LC)(total impurities): ≤ 0.8 %
  • Loss on drying (130 °C, 3 h): 10.5 - 12.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service