Skip to Content
Merck
All Photos(1)

Key Documents

1.01181

Supelco

Ammonium heptamolybdate tetrahydrate

(ammonium molybdate) powder extra pure

Synonym(s):

Ammonium heptamolybdate tetrahydrate, Ammonium molybdate, Hexammonium heptamolybdate 4-hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4 H2O
CAS Number:
Molecular Weight:
1235.86
MDL number:
UNSPSC Code:
12352302
EC Index Number:
234-722-4
NACRES:
NB.21
Pricing and availability is not currently available.

Quality Level

form

solid

pH

5.3 (20 °C, 50 g/L in H2O)

mp

90 °C (Elimination of water of crystallisation)

solubility

400 g/L

density

2.498 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

2-30°C

SMILES string

[Mo].[N+H4].[O-2].O

InChI

1S/Mo.H3N.H2O.O/h;1H3;1H2;/q;;;-2/p+1

InChI key

XCOMAVCHADVANQ-UHFFFAOYSA-O

Looking for similar products? Visit Product Comparison Guide

Application


  • A Novel Hydro-Thermal Synthesis of Nano-Structured Molybdenum-Iron Intermetallic Alloys at Relatively Low Temperatures.: This study explores the hydro-thermal synthesis of nano-structured molybdenum-iron intermetallic alloys using ammonium heptamolybdate tetrahydrate, highlighting its application in advanced material synthesis (El-Geassy et al., 2023).

  • Alternative Preparation of Improved NiMo-Alumina Deoxygenation Catalysts.: The research focuses on the use of ammonium heptamolybdate tetrahydrate in the preparation of NiMo-Alumina catalysts for deoxygenation reactions, emphasizing its role in catalytic chemistry (Priecel et al., 2020).

  • Chromatographic identification of "green capping agents" extracted from Nasturtium officinale (Brassicaceae) leaves for the synthesis of MoO(3) nanoparticles.: This paper discusses the synthesis of MoO3 nanoparticles using ammonium heptamolybdate tetrahydrate and green capping agents, showcasing its relevance in green chemistry and nanotechnology (Shaheen & Ahmad, 2020).

  • Preparation of h-MoO3 and α-MoO3 nanocrystals: comparative study on photocatalytic degradation of methylene blue under visible light irradiation.: The study investigates the photocatalytic properties of MoO3 nanocrystals synthesized from ammonium heptamolybdate tetrahydrate, highlighting its application in environmental chemistry for pollutant degradation (Chithambararaj et al., 2013).

  • Novel MoO2/carbon hierarchical nano/microcomposites: synthesis, characterization, solid state transformations and thiophene HDS activity.: This research focuses on the synthesis of MoO2/carbon composites using ammonium heptamolybdate tetrahydrate, emphasizing its catalytic applications in hydrodesulfurization (HDS) of thiophene (Avendaño et al., 2013).

Analysis Note

Assay (precipitative titration): ≥ 99.0 %
Identity: passes test
Chloride (Cl): ≤ 0.002 %
Phosphate, Arsenate, Silicate (as PO₄): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.02 %
Fe (Iron): ≤ 0.002 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service