Skip to Content
Merck
All Photos(1)

Key Documents

05-919

Sigma-Aldrich

Anti-T-Cell Receptor Antibody, clone C305

culture supernatant, clone C305, Upstate®

Synonym(s):

T-Cell receptor antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

C305, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
multiplexing: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TRDD3(28523)

Specificity

Jurkat T-Cell Receptor

Immunogen

Jurkat T cells

Application

Anti-T-Cell Receptor Antibody, clone C305 detects level of T-Cell Receptor & has been published & validated for use in Mplex & IP.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

routinely used in bioassays to activate T cell receptor signaling cascades in Jurkat cells

Physical form

Concentrated culture supernatant

Storage and Stability

stable 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Lin et al.
The Journal of cell biology, 162(4), 673-682 (2003-08-13)
CD148 is a receptor-like protein tyrosine phosphatase up-regulated on T cells after T cell receptor (TCR) stimulation. To examine the physiologic role of CD148 in TCR signaling, we used an inducible CD148-expressing Jurkat T cell clone. Expression of CD148 inhibits
A Weiss et al.
The Journal of experimental medicine, 160(5), 1284-1299 (1984-11-01)
The association between T3 and the T cell antigen receptor was examined using the T3 bearing T cell leukemic line Jurkat. A monoclonal antibody, C305, was produced, which reacted with idiotypic-like determinants expressed on Jurkat. The molecule with which this
Nadia Martinez-Martin et al.
Nature communications, 7, 11473-11473 (2016-05-06)
Viruses encode secreted and cell-surface expressed proteins essential to modulate host immune defenses and establish productive infections. However, to date there has been no systematic study of the extracellular interactome of any human virus. Here we utilize the E3 proteins
Pussadee Paensuwan et al.
Journal of immunology (Baltimore, Md. : 1950), 196(1), 448-458 (2015-11-22)
Ligand binding to the TCR causes a conformational change at the CD3 subunits to expose the CD3ε cytoplasmic proline-rich sequence (PRS). It was suggested that the PRS is important for TCR signaling and T cell activation. It has been shown
Tim Kükenshöner et al.
Journal of molecular biology, 429(9), 1364-1380 (2017-03-30)
The binding of Src-homology 2 (SH2) domains to phosphotyrosine (pY) sites is critical for the autoinhibition and substrate recognition of the eight Src family kinases (SFKs). The high sequence conservation of the 120 human SH2 domains poses a significant challenge

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service