Skip to Content
Merck
All Photos(5)

Documents

05-369

Sigma-Aldrich

Anti-Na+/K+ ATPase α-1 Antibody, clone C464.6

clone C464.6, Upstate®, from mouse

Synonym(s):

ATPase, Na+/K+ transporting, alpha 3 polypeptide, Na(+)/K(+) ATPase alpha(III) subunit, Na(+)/K(+) ATPase alpha-3 subunit, Na+/K+ -ATPase alpha 3 subunit, Na+/K+ ATPase 3, Sodium pump subunit alpha-3, dystonia 12, sodium pump 3, sodium-potassium-ATPase,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C464.6, monoclonal

species reactivity

rabbit, monkey, sheep, mouse, Xenopus, human, pig, canine, rat

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ATP1A3(478)

General description

Na+/K+-ATPase (also known as the Na+/K+ pump or sodium-potassium pump) is an enzyme located in the plasma membrane (specifically an electrogenic transmembrane ATPase). It is found in the plasma membrane of virtually every human cell and is common to all cellular life. It helps maintain cell potential and regulate cellular volume.
In order to maintain the cell potential, cells must keep a low concentration of sodium ions and high levels of potassium ions within the cell (intracellular). Outside cells (extracellular), there are high concentrations of sodium and low concentrations of potassium, so diffusion occurs through ion channels in the plasma membrane. In order to keep the appropriate concentrations, the sodium-potassium pump pumps sodium out and potassium in through active transport.

Specificity

This antibody recognizes the α-1 subunit isoform of Na+/K+ ATPase.

Immunogen

Purified Na+/K+ ATPase isolated from membrane fractions of rabbit kidney outer medulla.

Application

Detect Na+/K+ ATPase α-1 (sodium/potassium ATPase subunit alpha 1) using this Anti-Na+/K+ ATPase α-1 Antibody, clone C464.6 validated for use in IC, IH, IP & WB.
Immunoprecipitation:
2 µg of a previous lot immuno-precipitated the α-1 subunit from rat brain microsomal preparations.

Immunohistochemistry:
1-10 µg/mL of a previous lot of this antibody was used in immunohistochemistry on ice-cold methanol-fixed tissue.

1. Do not boil microsomes. Warming to 37ºC for 15 minutes is recommended.
2. Microsomes from some tissues may contain sodium pump subunits which are below the level of detection by this antibody.
3. Microsomes derived from tissues which are more difficult to prepare, such as smooth and skeletal muscle, frequently show non-specific cross-reactivity.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Quality

Routinely evaluated in western blot on rat brain microsomal preparations.

Western Blot Analysis:
0.05-0.1 µg/mL of this lot detected the α-1 subunit in 20 µg of rat brain microsomal preparation (Catalog # 12-144). To reduce background, wash the blot with 0.05% Tween-20 in PBS for 15 minutes.

Target description

112 kDa

Physical form

Format: Purified
Protein G Chromatography
Purified mouse monoclonal IgG1κ in buffer containing 0.02 M phosphate buffer, pH 7.6, 0.25 M NaCl, 0.1% sodium azide. Frozen at
-20ºC.
Produced from a culture supernatant of a hybridoma produced by fusing Ag8 mouse myeloma cells with immunized Balb/c splenocytes. Clone C464.6 also called 6H.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Alpha 1 mRNA is present in all cell types examined.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reorganization of lipid rafts during capacitation of human sperm.
Cross, NL
Biology of reproduction null
Isoforms of the Na,K-ATPase are present in both axons and dendrites of hippocampal neurons in culture.
Pietrini, G, et al.
Proceedings of the National Academy of Sciences of the USA, 89, 8414-8418 (1992)
N B Illarionova et al.
Neuroscience, 168(4), 915-925 (2009-12-08)
The water channel aquaporin 4 (AQP4) is abundantly expressed in astrocytes and provides a mechanism by which water permeability of the plasma membrane can be regulated. Astrocytes play a key role in the clearance of both potassium (K(+)) and glutamate
Renal phenotype of UT-A urea transporter knockout mice.
Fenton, RA; Flynn, A; Shodeinde, A; Smith, CP; Schnermann, J; Knepper, MA
Journal of the American Society of Nephrology null
Vasopressin regulation of the renal UT-A3 urea transporter.
Stewart, GS; Thistlethwaite, A; Lees, H; Cooper, GJ; Smith, C
American Journal of Physiology: Renal Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service