Skip to Content
Merck
All Photos(1)

Documents

V-017

Supelco

Nicotinic acid (Vitamin B3) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Nicotinic acid, Vitamin B3, 3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

236-239 °C (lit.)

density

0.791 g/cm3 at 20 °C

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic acid, also known as niacin, is a water-soluble vitamin of the B complex and essential human nutrient. Niacin deficiency, often the result of impaired nutritional status associated with conditions ranging from malnutrition to chronic alcoholism, is monitored in patient plasma samples by HPLC. This Certified Spiking Solution® is suitable for use as a starting material in the preparation of linearity standards, calibrators or controls in a variety of testing applications from diagnostic testing of niacin deficiency by HPLC to quality control testing of niacin-based pharmaceutical preparations, dietary supplements, or fortified foods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niacin therapy lives for another day-maybe?
Carl J Lavie et al.
Journal of the American College of Cardiology, 61(21), 2197-2198 (2013-04-02)
Wanda C Lakey et al.
Journal of clinical lipidology, 7(2), 178-181 (2013-02-19)
Multiple cholesterol-reducing therapies have been shown to induce the regression of tendon xanthoma in patients with familial hypercholesterolemia. We present 3 cases of adverse reactions in Achilles tendon xanthomas after the addition of niacin and bile acid sequestrants to ongoing
Drugs for hypertriglyceridemia.
The Medical letter on drugs and therapeutics, 55(1411), 17-19 (2013-03-08)
Rachel L Edwards et al.
Infection and immunity, 81(3), 945-955 (2013-01-16)
In response to environmental fluctuations or stresses, bacteria can activate transcriptional and phenotypic programs to coordinate an adaptive response. The intracellular pathogen Legionella pneumophila converts from a noninfectious replicative form to an infectious transmissive form when the bacterium encounters alterations
N Nakajima et al.
Clinical toxicology (Philadelphia, Pa.), 51(5), 451-454 (2013-05-07)
Erythromelalgia is a rare disorder characterized by reddening, severe burning pain, and swelling of the extremities. Food poisoning by Clitocybe acromelalga, a poisonous mushroom, is known to induce erythromelalgia; however, its treatment protocol remains unclear. We describe here three cases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service