Skip to Content
Merck
All Photos(1)

Key Documents

S-021

Supelco

Sertraline hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18Cl3N
CAS Number:
Molecular Weight:
342.69
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13

InChI

1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1

InChI key

BLFQGGGGFNSJKA-XHXSRVRCSA-N

Gene Information

human ... SLC6A4(6532)

General description

Sertraline, marketed as the brand drug Zoloft®, is an SSRI antidepressant used in the treatment of major depression as well as obsessive-compulsive, panic, and social anxiety disorders. This Certified Snap-N-Spike® Solution is applicable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Zoloft is a registered trademark of Pfizer, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 1 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andre R Brunoni et al.
JAMA psychiatry, 70(4), 383-391 (2013-02-08)
Transcranial direct current stimulation (tDCS) trials for major depressive disorder (MDD) have shown positive but mixed results. To assess the combined safety and efficacy of tDCS vs a common pharmacological treatment (sertraline hydrochloride, 50 mg/d). Double-blind, controlled trial. Participants were
Quang A Le et al.
Health and quality of life outcomes, 11, 59-59 (2013-04-17)
To determine the minimal clinically important difference (MCID) for the health-utility measures EuroQol-5 dimensions (EQ-5D) and Quality of Well Being Self-Administered (QWB-SA) Scale in PTSD patients. Two hundred patients aged 18 to 65 years with PTSD enrolled in a doubly
Latif Moradveisi et al.
The British journal of psychiatry : the journal of mental science, 202(3), 204-211 (2013-02-09)
Behavioural activation might be a viable alternative to antidepressant medication for major depressive disorder. To compare the effectiveness of behavioural activation and treatment as usual (TAU, antidepressant medication) for major depressive disorder in routine clinical practice in Iran. Patients with
Alastair J Flint et al.
BMC psychiatry, 13, 38-38 (2013-01-29)
Psychotic depression (PD) is a severe disabling disorder with considerable morbidity and mortality. Electroconvulsive therapy and pharmacotherapy are each efficacious in the treatment of PD. Expert guidelines recommend the combination of antidepressant and antipsychotic medications in the acute pharmacologic treatment
Yvette I Sheline et al.
The American journal of psychiatry, 169(11), 1185-1193 (2013-03-28)
In order to assess the effect of gray matter volumes and cortical thickness on antidepressant treatment response in late-life depression, the authors examined the relationship between brain regions identified a priori and Montgomery-Åsberg Depression Rating Scale (MADRS) scores over the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service