Skip to Content
Merck
All Photos(1)

Key Documents

P-070

Supelco

Progesterone-D9 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21O2D9
CAS Number:
Molecular Weight:
323.52
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1=C2C([2H])([2H])CC3C(CCC4(C)C3CC[C@]4([2H])C(=O)C([2H])([2H])[2H])C2(C)CC([2H])([2H])C1=O

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1/i1D3,4D2,8D2,12D,17D

InChI key

RJKFOVLPORLFTN-PQIPVKAESA-N

General description

Progesterone, a steroid hormone of the progestogen class, is used for hormone replacement therapy. This internal standard is applicable for progesterone testing or isotope dilution methods by GC/MS or LC/MS for endocrinology, female health testing or clinical chemistry.

Application


  • Progesterone-D₉ solution for hormone research: Progesterone-D₉ is extensively used as an internal standard in pharmacokinetic studies to measure progesterone levels in biological samples using LC-MS/MS, enhancing the accuracy of hormonal assays critical for understanding progesterone′s role in various physiological processes (Sasaki et al., 2015).

  • Stable isotope labeled progesterone for bioanalysis: As a stable isotope-labeled compound, Progesterone-D₉ is integral in mass spectrometry, providing a reliable standard for quantifying progesterone in complex biochemical assays, thus facilitating detailed studies on steroid metabolism and its implications.

  • Deuterated progesterone for mass spectrometry: The application of Progesterone-D₉ in mass spectrometry enhances the precision of detecting and quantifying minute quantities of progesterone and its metabolites, which is crucial for drug development and therapeutic monitoring.

  • Progesterone-D₉ in steroid metabolism studies: Utilized in research to understand the metabolic pathways of progesterone, Progesterone-D₉ helps in tracing the transformation and interaction of this hormone within the body, supporting advancements in reproductive health and disease treatment.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service