Skip to Content
Merck
All Photos(1)

Key Documents

M-152

Supelco

Mitragynine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N2O4
CAS Number:
Molecular Weight:
398.50
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(OC)/C([C@H]1C[C@@](N(C[C@H]1CC)CC2)([H])C3=C2C(C(OC)=CC=C4)=C4N3)=C/OC

InChI

1S/C23H30N2O4/c1-5-14-12-25-10-9-15-21-18(7-6-8-20(21)28-3)24-22(15)19(25)11-16(14)17(13-27-2)23(26)29-4/h6-8,13-14,16,19,24H,5,9-12H2,1-4H3/b17-13+/t14-,16+,19+/m1/s1

InChI key

LELBFTMXCIIKKX-QVRQZEMUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mitragynine, commonly known as Kratom, is the most abundant alkaloid in the plant Mitragynine speciosa. Sold over the internet as herbal plant extracts, Kratom is noted for its psychoactive, stimulant, and sedative effects depending on the dosage level. According to DEA′s Drugs and Chemicals of Concern page, mitragynine use can lead to addiction with adverse effects ranging from anorexia and weight loss to hallucinations and delusion.

Application



  • Pharmacokinetic Interaction of Kratom and Cannabidiol: A study analyzed the pharmacokinetic interactions between mitragynine and cannabidiol in male rats, providing critical insights for co-administration of these compounds (Berthold et al., 2024).


  • Enhancement of Low-Dose Doxorubicin Cytotoxicity by Kratom Extracts: Research investigated whether kratom extracts enhance the cytotoxic effects of low-dose doxorubicin against lung cancer cells, suggesting a potential use of mitragynine in cancer therapy (Bayu et al., 2024).


  • Extraction and Detection of Mitragynine: A technique using high-performance liquid chromatography for the extraction and detection of mitragynine from Kratom leaves was developed, enhancing the accuracy and efficiency of mitragynine isolation for research purposes (Ng and Ha, 2024).


  • Amelioration of Mitragynine Withdrawal Symptoms: A study explored the potential of an epigenetic mechanism to ameliorate withdrawal behavior and cognitive impairments caused by mitragynine, contributing to better management strategies for opioid withdrawal symptoms (Yunusa et al., 2024).


  • Imaging Mass Spectrometry Analysis of Mitragynine: Research utilized MALDI imaging mass spectrometry to study the distribution of Mitragyna speciosa alkaloids, including mitragynine, in dosed rat brain tissue, advancing the understanding of its biochemical behavior (Liang et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beng-Siang Khor et al.
PloS one, 6(12), e28340-e28340 (2011-12-30)
A major obstacle in treating drug addiction is the severity of opiate withdrawal syndrome, which can lead to unwanted relapse. Mitragynine is the major alkaloid compound found in leaves of Mitragyna speciosa, a plant widely used by opiate addicts to
Anika A Philipp et al.
Analytical and bioanalytical chemistry, 399(8), 2747-2753 (2011-01-21)
Mitragyna speciosa (Kratom) is currently used as a drug of abuse. When monitoring its abuse in urine, several alkaloids and their metabolites must be considered. In former studies, mitragynine (MG), its diastereomer speciogynine (SG), and paynantheine and their metabolites could
Xuefeng Sun et al.
Chemistry, an Asian journal, 6(8), 2158-2165 (2011-06-15)
O-Trimethylsilyl (TMS)-protected diphenylprolinol-catalyzed Michael addition of a functionalized alkylidene malonate and n-butanal affords an aldehyde. This adduct can serve as the common intermediate for the assembly of secologanin tryptamine and dopamine alkaloids; this is demonstrated by the total syntheses of
Torsten Arndt et al.
Forensic science international, 208(1-3), 47-52 (2010-11-30)
A drug and alcohol withdrawal rehabilitation centre requested an analysis for "Krypton" in urine of a former opiate-addictive woman. She showed an altered clinical picture and behaviour with miosis, itchiness, agitation, and moderate euphoria after 3 months of until than
Ee Lin Lim et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(2), 812-824 (2013-01-01)
CYP450 enzymes are key determinants in drug toxicities, reduced pharmacological effect and adverse drug reactions. Mitragynine, an euphoric compound was evaluated for its effects on the expression of mRNAs encoding CYP1A2, CYP2D6 and CYP3A4 and protein expression and resultant enzymatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service