Skip to Content
Merck
All Photos(1)

Documents

D-039

Supelco

N-Desethylamodiaquine dihydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18ClN3O·2HCl
CAS Number:
Molecular Weight:
400.73
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC=C(C(NC2=CC=C(O)C(CNCC)=C2)=CC=N3)C3=C1.Cl.Cl

InChI

1S/C18H18ClN3O.2ClH/c1-2-20-11-12-9-14(4-6-18(12)23)22-16-7-8-21-17-10-13(19)3-5-15(16)17;;/h3-10,20,23H,2,11H2,1H3,(H,21,22);2*1H

InChI key

BCYBRFGUODDEGH-UHFFFAOYSA-N

General description

N-Desethylamodiaquine is an active primary blood metabolite of the antimalarial drug amodiaquine. This certified solution standard is applicable for use in clinical toxicology by LC-MS/MS or GC/MS. Amodiaquine, sold under trade names Camoquin and Flavoquine, is used as an anti-inflammatory agent and as a treatment for malaria.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Sarrassat et al.
Acta tropica, 110(1), 1-6 (2008-12-06)
The deployment of Artemisinin-based Combination Therapy for treating uncomplicated malaria poses problems in the patient compliance to these new treatments. The aim of our study was to investigate the relationship between compliance to 3 days treatment with Artesunate plus Amodiaquine
Anupkumar R Anvikar et al.
The Indian journal of medical research, 135(4), 494-499 (2012-06-06)
In vitro assays are an important tool to assess baseline sensitivity and monitor the drug response of Plasmodium falciparum over time and place and, therefore, can provide background information for the development and evaluation of drug policies. This study was
George O Adjei et al.
Malaria journal, 11, 420-420 (2012-12-19)
Several anti-malarial drugs are associated with adverse cardiovascular effects. These effects may be exacerbated when different anti-malarials are used in combination. There has been no report yet on the potential cardiac effects of the combination artesunate-amodiaquine. Electrocardiographic (ECG) intervals in
V Sinou et al.
European journal of drug metabolism and pharmacokinetics, 34(3-4), 133-142 (2010-02-20)
A new fixed-dose combination of artesunate (AS) plus amodiaquine (AQ) (TRIMALACT) was recently developed for the treatment of uncomplicated falciparum malaria. The originality of this combination lies in its galenic formulation which consists of a three-layer tablet with two layers
Adriana Pabón et al.
Experimental parasitology, 127(1), 222-227 (2010-08-26)
The effect of 16 alpha-acetoxy-26-hydroxycholest-4-ene-3,22-dione (SN-1) isolated from Solanum nudum Dunal (a Solanaceae traditionally used for treating fever in Colombia) on Plasmodium falciparum erythrocyte stages and its in vitro antiplasmodial activity when combined with the following conventional drugs was studied:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service