Skip to Content
Merck
All Photos(1)

Documents

H-902

Supelco

α-Hydroxymidazolam solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13ClFN3O
CAS Number:
Molecular Weight:
341.77
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OCc1ncc2CN=C(c3ccccc3F)c4cc(Cl)ccc4-n12

InChI

1S/C18H13ClFN3O/c19-11-5-6-16-14(7-11)18(13-3-1-2-4-15(13)20)22-9-12-8-21-17(10-24)23(12)16/h1-8,24H,9-10H2

InChI key

QHSMEGADRFZVNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Hydroxymidazolam is an active, major metabolite in urine and blood of the benzodiazepine, midazolam. Midazolam is sold as Dormicum, Hypnovel®, and Versed as a sedative and treatment for insomnia and seizures. This Certified Spiking Solution® is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.

Application


  • α-Hydroxymidazolam solution in pharmacokinetic studies: The application of α-Hydroxymidazolam solution is crucial in pharmacokinetic research to study the metabolism of midazolam, particularly its phase I metabolic pathways, providing essential data on the drug′s biotransformation and clearance rates in humans (Bourget et al., 1996).

  • α-Hydroxymidazolam as a benzodiazepine metabolite standard: Utilized as a benzodiazepine metabolite standard, α-Hydroxymidazolam aids in the accurate measurement of midazolam and its metabolites in plasma using high-performance liquid chromatography, essential for ensuring dosing efficacy and safety in clinical settings (Lee and Charles, 1996).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Hypnovel is a registered trademark of Hoffman-LaRoche & Co. AG
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruut Kaartama et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(19), 1668-1676 (2011-05-03)
A sensitive and selective gas chromatographic mass spectrometric method for the determination of midazolam and its biologically active metabolite, 1-hydroxymidazolam, in rabbit plasma has been developed and validated. Sample preparation includes mixed-mode solid-phase extraction and derivatization with silylating reagents. Midazolam-d4
High-throughput analysis of in vitro cytochrome p450 inhibition samples using mass spectrometry coupled with an integrated liquid chromatography/autosampler system.
Ann Brown et al.
Rapid communications in mass spectrometry : RCM, 24(8), 1207-1210 (2010-03-20)
Kai-Ming Duan et al.
Journal of clinical pharmacology, 52(6), 940-946 (2011-06-18)
The aims of this study were to investigate the effect of quercetin on CYP3A activity in vivo. An open, randomized, 2-period crossover experiment was performed in 18 healthy male volunteers. Genotyped data were available from a total of 165 participants.
J Atzrodt et al.
Bioorganic & medicinal chemistry, 20(18), 5658-5667 (2012-08-15)
The syntheses of stable isotope labelled internal standards of important CYP-isoform selective probes, like testosterone 1, diclofenac 3, midazolam 5, and dextromethorphan 7, as well as their corresponding hydroxylated metabolites 6β-hydroxytestosterone 2, 4'-hydroxydiclofenac 4, 1'-hydroxymidazolam 6 and dextrorphan 8 are
Toshiyuki Iwasaki et al.
Pediatrics international : official journal of the Japan Pediatric Society, 52(4), 513-519 (2009-12-17)
The aim of the present study was to determine an index to evaluate the efficacy and safety of midazolam (MDZ) to treat status epilepticus (SE). An original system was therefore developed to measure blood concentrations of MDZ and 1-hydroxymidazolam (1-OHMDZ)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service