Skip to Content
Merck
All Photos(1)

Key Documents

A-018

Supelco

(±)-Amphetamine-d8 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-Amphetamine-d8 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D8H5N
CAS Number:
Molecular Weight:
143.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

NC(C([2H])([2H])[2H])CC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]

InChI

1S/C9H13N/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/i1D3,2D,3D,4D,5D,6D

InChI key

KWTSXDURSIMDCE-JGUCLWPXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amphetamine, a stimulant of the phenethylamine class, is found in ADHD-treatment drugs such as Adderall®, Vyvanse®, and ProCentra®. Because of its stimulant effect, amphetamine is used recreationally as a performance-enhancer and illicit drug. This stable-labeled internal standard is suitable for quantitation of amphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or sports testing.

Application


  • Quantification of amphetamine plasma concentrations: A study details a method using gas chromatography coupled to mass spectrometry (GC-MS) for quantifying amphetamine concentrations in plasma. This technique utilizes deuterated amphetamine, such as Amphetamine-d₈, as an internal standard to ensure accuracy and reliability in measurements, crucial for pharmacokinetic analyses and drug metabolism studies in pharmaceutical research (Pizarro et al., 1999).


Legal Information

Adderall is a registered trademark of Richwood Pharmaceutical Company, Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
ProCentra is a registered trademark of Outlook Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyvanse is a registered trademark of Shire, LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service