Skip to Content
Merck
All Photos(1)

Key Documents

E-018

Supelco

(±)-N-Ethylamphetamine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N
CAS Number:
Molecular Weight:
163.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CCNC(C)Cc1ccccc1

InChI

1S/C11H17N/c1-3-12-10(2)9-11-7-5-4-6-8-11/h4-8,10,12H,3,9H2,1-2H3

InChI key

YAGBSNMZQKEFCO-UHFFFAOYSA-N

General description

N-Ethylamphetamine is a stimulant drug of the phenethylamine and amphetamine classes. This certified solution standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from sports testing and clinical toxicology to forensic analysis and urine drug testing. N-Ethylamphetamine, sold as the pharmaceutical appetite-suppressant Apetinil, is also abused as a recreational drug with a stimulant effect similar to that of its amphetamine and methamphetamine analogs.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriele Cruciani et al.
Journal of medicinal chemistry, 48(22), 6970-6979 (2005-10-28)
Identification of metabolic biotransformations can significantly affect the drug discovery process. Since bioavailability, activity, toxicity, distribution, and final elimination all depend on metabolic biotransformations, it would be extremely advantageous if this information could be produced early in the discovery phase.
F T Delbeke et al.
Arzneimittel-Forschung, 36(9), 1413-1416 (1986-09-01)
The urinary excretion of etilamfetamine (ethylamphetamine) and its major metabolite amphetamine in humans was followed over a period of several days after the oral administration of two formulations. The excretion of both substances was affected by urinary pH. Excretion peaks
K Matsushima et al.
Nihon hoigaku zasshi = The Japanese journal of legal medicine, 52(1), 19-26 (1998-05-20)
Characterization of optical activity and simultaneous analysis of racemic ethylamphetamine (EAMP) and its metabolites, as well as the urinary excretion of the optical isomers, were examined in rats by high-performance liquid chromatography (HPLC). Analysis of the optical isomers of EAMP
A Dasgupta et al.
Journal of forensic sciences, 43(3), 636-640 (1998-06-03)
Phenmetrazine is a central nervous system stimulant currently used as an anorectic agent. The drug is abused and is reported to cause death from overdose. We describe a new derivatization method for phenmetrazine using 4-carbethoxyhexafluorobutyryl chloride. Quantitation of urinary phenmetrazine
Y Makino et al.
Forensic science international, 41(1-2), 83-91 (1989-04-01)
Ethylamphetamine and two major metabolites in urine, obtained after intraperitoneal administration of the drug to rats, have been isolated by gas chromatography-mass spectrometry. We have also synthesized an authentic sample of p-hydroxy-N-ethylamphetamine by a simple method. In rat, the metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service