Skip to Content
Merck
All Photos(1)

Documents

880124X

Avanti

DSPE-PEG(2000) Folate

Avanti Research - A Croda Brand 880124X

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-2000] (ammonium salt):methanol (9:1)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (880124X-10mg)
pkg of 1 × 1 mL (880124X-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880124X

concentration

1 mg/mL (880124X-1mg)
10 mg/mL (880124X-10mg)

shipped in

dry ice

storage temp.

−20°C

General description

DSPE-PEG(2000) Folate, also known as 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-2000], is a functionalized PEG lipid. This lipid is added into liposomes as a targeting ligand for folate receptors, which acts as cellular surface markers for numerous solid tumors and myeloid leukemias. Folate targeted drug delivery is used an effective therapy for the treatment and imaging of various cancers and inflammatory diseases. Folate conjugates with low molecular weight and high binding affinity for folate receptors (FR) on cell surface deliver a variety of molecular complexes such as protein toxins, immune stimulants, chemotherapeutic agents, liposomes, nanoparticles, and imaging agents to pathologic cells without affecting normal tissues.

Application

DSPE-PEG(2000) Folate(1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-2000]) may be used in preparation of folate-conjugated liposomes and folate-modified microvesicles (MVs).

Packaging

5 mL Clear Glass Sealed Ampule (880124X-10mg)
5 mL Clear Glass Sealed Ampule (880124X-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Wei Deng et al.
Nature communications, 9(1), 2713-2713 (2018-07-15)
Liposomes have been well established as an effective drug delivery system, due to simplicity of their preparation and unique characteristics. However conventional liposomes are unsuitable for the on-demand content release, which limits their therapeutic utility. Here we report X-ray-triggerable liposomes
Controlled gene and drug release from a liposomal delivery platform triggered by X-ray radiation
Deng W, et al.
Nature Communications, 9(1) (2018)
Wei Zhang et al.
ACS nano, 11(1), 277-290 (2016-12-23)
Cell-derived microvesicles (MVs), which are biogenic nanosized membrane-bound vesicles that convey bioactive molecules between cells, have recently received attention for use as natural therapeutic platforms. However, the medical applications of MV-based delivery platforms are limited by the lack of effective
Magnetic and folate functionalization enables rapid isolation and enhanced tumor-targeting of cell-derived microvesicles
Zhang W, et al.
ACS Nano, 11(1), 277-290 (2017)
Andrew R Hilgenbrink et al.
Journal of pharmaceutical sciences, 94(10), 2135-2146 (2005-09-02)
Folate targeted drug delivery has emerged as an alternative therapy for the treatment and imaging of many cancers and inflammatory diseases. Due to its small molecular size and high binding affinity for cell surface folate receptors (FR), folate conjugates have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service