Skip to Content
Merck
All Photos(1)

Key Documents

880001O

Avanti

Chol-PEG600

Avanti Research - A Croda Brand 880001O

Synonym(s):

Cholesterol-(polyethylene glycol-600)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 25 mg (880001O-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880001O

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C29H50O2/c1-20(2)7-6-8-21(3)25-11-12-26-24-10-9-22-19-23(31-18-17-30)13-15-28(22,4)27(24)14-16-29(25,26)5/h9,20-21,23-27,30H,6-8,10-19H2,1-5H3

InChI key

NPYXBHZQVNRAOF-UHFFFAOYSA-N

General description

Cholesterol-(polyethylene glycol-600) (Chol-PEG600) is a nonionic amphipathic lipid and a cholesterol derivative. It is a water soluble, non-homogeneous compound containing hydrophobic cholesterol and covalently bound hydrophilic PEG.

Biochem/physiol Actions

PEG-cholesterol is widely used for cationic DNA transfection studies, and preparation of liposomes for encapsulated drug delivery.

Packaging

5 mL Clear Glass Sealed Ampule (880001O-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (ethylene glycol)-cholesterol inhibits L-type Ca2+ channel currents and augments voltage-dependent inactivation in A7r5 cells
Ochi, R, et al.
PLoS ONE, 9(9) (2014)
Poly (ethylene glycol)-cholesterol inhibits L-type Ca2+ channel currents and augments voltage-dependent inactivation in A7r5 cells
Ochi, R, et al.
Testing, 9(9) (2014)
Rikuo Ochi et al.
PloS one, 9(9), e107049-e107049 (2014-09-10)
Cholesterol distributes at a high density in the membrane lipid raft and modulates ion channel currents. Poly(ethylene glycol) cholesteryl ether (PEG-cholesterol) is a nonionic amphipathic lipid consisting of lipophilic cholesterol and covalently bound hydrophilic PEG. PEG-cholesterol is used to formulate
CHOBIMALT: a cholesterol-based detergent
Howell S, et al.
Biochemistry, 49(44), 9572-9583 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service