Skip to Content
Merck
All Photos(1)

Key Documents

860525P

Avanti

C24:1 Ceramide (d18:1/24:1(15Z))

Avanti Research - A Croda Brand

Synonym(s):

N-nervonoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H81NO3
CAS Number:
Molecular Weight:
648.10
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860525P-10mg)
pkg of 1 × 25 mg (860525P-25mg)
pkg of 1 × 5 mg (860525P-5mg)
pkg of 1 × 50 mg (860525P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C42H81NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-42(46)43-40(39-44)41(45)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h17-18,35,37,40-41,44-45H,3-16,19-34,36,38-39H2,1-2H3,(H,43,46)/b18-17-,37-35+/t40-,41+/m0/s1

InChI key

VJSBNBBOSZJDKB-KPEYJIHVSA-N

General description

Ceramides comprise spingolipids with acyl chains of C14 to C24 saturated and monounsaturated fatty acids. They are present in most of the mammalian tissues.

Application

C24:1 Ceramide (d18:1/24:1(15Z)) has been used as an internal stand in liquid chromatography-mass spectrometer. It is also suitable for use as a standard in high-performance liquid chromatography/tandem mass spectrometry.

Biochem/physiol Actions

Ceramide (d18:1/24:1[15Z]) levels are elevated inprimary open angle glaucoma (POAG). It is a biomarker for the human arterial tissue and plasma major adverse cardiac and cerebrovascular events (MACE). In general, ceramides are key second messengers in sphingomyelin pathway and modulate the pathophysiology of metabolic diseases.

Packaging

5 mL Amber Glass Screw Cap Vial (860525P-10mg)
5 mL Amber Glass Screw Cap Vial (860525P-25mg)
5 mL Amber Glass Screw Cap Vial (860525P-50mg)
5 mL Amber Glass Screw Cap Vial (860525P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human trabecular meshwork sphingolipid and ceramide profiles and potential latent fungal commensalism
Aljohani AJ, et al.
Investigative Ophthalmology & Visual Science, 55(6), 3413-3422 (2014)
Atypical surface behavior of ceramides with nonhydroxy and 2-hydroxy very long-chain (C28-C32) PUFAs
Penalva DA, et al.
Biochimica et Biophysica Acta - Biomembranes, 1838(3), 731-738 (2014)
Plasma ceramides as prognostic biomarkers and their arterial and myocardial tissue correlates in acute myocardial infarction
de Carvalho LP, et al.
JACC. Basic to translational science, 3(2), 163-175 (2018)
Hypothalamic AMPK-ER stress-JNK1 axis mediates the central actions of thyroid hormones on energy balance
Martinez-Sanchez N, et al.
Cell Metabolism, 26(1), 212-229 (2017)
Increased ceramide content and NFkappaB signaling may contribute to the attenuation of anabolic signaling after resistance exercise in aged males
Rivas DA, et al.
Journal of Applied Physiology, 113(11), 1727-1736 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service