Skip to Content
Merck
All Photos(2)

Key Documents

855575P

Avanti

14:0 Lyso PC

1-myristoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-tetradecanoyl-sn-glycero-3-phosphocholine; PC(14:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H46NO7P
CAS Number:
Molecular Weight:
467.58
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 1 g (855575P-1g)
pkg of 1 × 200 mg (855575P-200mg)
pkg of 1 × 25 mg (855575P-25mg)
pkg of 1 × 500 mg (855575P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855575P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCC)=O

InChI

1S/C22H46NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-22(25)28-19-21(24)20-30-31(26,27)29-18-17-23(2,3)4/h21,24H,5-20H2,1-4H3/t21-/m1/s1

InChI key

VXUOFDJKYGDUJI-OAQYLSRUSA-N

Application

14:0 Lyso PC has been used to study its role in nerve growth factor (NGF)-induced mitogen-activated protein kinase (MAPK) and serine/threonine-protein kinase akt-1 signals in rat pheochromocytoma PC12 cells.[1]
14:0 Lyso PC may be used:
  • as a component of detergent micelle to analyze the secondary structure of the purified twin-arginine translocation (Tat) membrane protein (TatCd) by circular dichroism (CD)[2]
  • for the synthesis of three monofluorinated dimyristoylphosphatidylcholines (F-DMPC′s), such as 2F-DMPC, 12F-DMPC and 14F-DMPC[3]
  • as a component of dry lipid film for the synthesis of palladium (Pd)-pyrolipid theranostic nanoparticles (PdPL-NPs) and manganese-pyrolipid surface-enhanced Raman scattering (SERS) NPs (MnPL-NPs)[4]

Packaging

20 mL Clear Glass Screw Cap Vial (855575P-1g)
20 mL Clear Glass Screw Cap Vial (855575P-500mg)
5 mL Amber Glass Screw Cap Vial (855575P-200mg)
5 mL Amber Glass Screw Cap Vial (855575P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of the effect of fluorination on the property of monofluorinated dimyristoylphosphatidylcholines
Gagnon MC, et al.
Organic & Biomolecular Chemistry, 12(28), 5126-5135 (2014)
Lysophosphatidylcholine enhances NGF-induced MAPK and Akt signals through the extracellular domain of TrkA in PC12 cells
Itakura A, et al.
FEBS Open Bio, 3, 243-251 (2013)
Structure analysis of the membrane protein TatCd from the Tat system of B. subtilis by circular dichroism
Nolandt OV, et al.
Biochimica et Biophysica Acta - Biomembranes, 1788(10), 2238-2244 (2009)
Nano-enabled SERS reporting photosensitizers
Farhadi A, et al.
Theranostics, 5(5), 469-469 (2015)
Reinhard Zeisig et al.
Cancer gene therapy, 10(4), 302-311 (2003-04-08)
To improve liposomal gene transfer we investigated the influence of membrane-interacting alkylphospholipids (APLs) on gene transfer efficiency in vitro and in vivo using the LacZ reporter gene and the cytosine deaminase (CD) suicide gene. Liposomes were first optimized concerning the

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service