Skip to Content
Merck
All Photos(3)

Documents

473936

Sigma-Aldrich

N-Methylformamide

99%

Synonym(s):

N-Formylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONHCH3
CAS Number:
Molecular Weight:
59.07
Beilstein:
1098352
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.432 (lit.)

bp

198-199 °C (lit.)

mp

−4 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)NC

InChI

1S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)

InChI key

ATHHXGZTWNVVOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methylformamide (NMF), a formamide derivative, is widely employed as an organic solvent. NMF molecule contains C=O and N-H groups, so it can act as a proton donor and acceptor. Its antitumor activities in mice have been assessed. Optical-heterodyne-detected optical Kerr effect (OHD-OKE) spectroscopic evaluation of NMF has been reported. Due to its important physical properties such as high dielectric constant, high solubility, low conductivity and amphiprotic nature, it is suitable for use as a separation medium in capillary electrophoresis studies.

Application

N-Methylformamide may be employed in the preparation of the following:
  • Li2S-P2S5 solid electrolytes (SE)
  • interlayer-condensed materials of protonated layered silicate (magadiite)
  • Li3PS4 solid electrolyte

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid-phase synthesis of a Li3PS4 solid electrolyte using N-methylformamide for all-solid-state lithium batteries.
Teragawa S, et al.
Journal of Material Chemistry A, 2(14), 5095-5099 (2014)
Femtosecond dynamics of hydrogen-bonding solvents. Formamide and N-methylformamide in acetonitrile, DMF, and water.
Chang JY and Castner Jr EW.
J. Chem. Phys. , 99(1), 113-125 (1993)
Preparation of Li2S-P2S5 solid electrolyte from N-methylformamide solution and application for all-solid-state lithium battery.
Teragawa S, et al.
Journal of Power Sources, 248, 939-942 (2014)
Interlayer condensation of protonated layered silicate magadiite through refluxing in N-methylformamide.
Asakura Y, et al.
Bulletin of the Chemical Society of Japan, 88(9), 1241-1249 (2015)
Jeppe S Spicker et al.
Toxicological sciences : an official journal of the Society of Toxicology, 102(2), 444-454 (2008-01-08)
A large number of databases are currently being implemented within toxicology aiming to integrate diverse biological data, such as clinical chemistry, expression, and other types of data. However, for these endeavors to be successful, tools for integration, visualization, and interpretation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service