Skip to Content
Merck
All Photos(1)

Key Documents

850340P

Avanti

13:0 PC

1,2-ditridecanoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

PC(13:0/13:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H68NO8P
CAS Number:
Molecular Weight:
649.88
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850340P-200mg)
pkg of 1 × 25 mg (850340P-25mg)
pkg of 1 × 500 mg (850340P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850340P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCC)=O)COC(CCCCCCCCCCCC)=O)=O

InChI

1S/C34H68NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-33(36)40-30-32(31-42-44(38,39)41-29-28-35(3,4)5)43-34(37)27-25-23-21-19-17-15-13-11-9-7-2/h32H,6-31H2,1-5H3/t32-/m1/s1

InChI key

RLNYFMQMIIHWFV-JGCGQSQUSA-N

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes. It is also an important component of the mucosal layer of the colon.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

13:0 PC is suitable for use:
  • in the preparation of the giant unilamellar vesicles (GUVs)
  • in the preparation of lipid monolayers
  • as an internal standard for the extraction of total lipids from biological samples

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance. It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.

Packaging

20 mL Clear Glass Screw Cap Vial (850340P-500mg)
5 mL Amber Glass Screw Cap Vial (850340P-200mg)
5 mL Amber Glass Screw Cap Vial (850340P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two-photon fluorescence microscopy observation of shape changes at the phase transition in phospholipid giant unilamellar vesicles
Bagatolli LA and Gratton E
Biophysical Journal, 77(4), 2090-2101 (1999)
The Membranes of Cells (2016)
Integrative Medicine - E-Book (2017)
Oxidized phospholipids are more potent antagonists of lipopolysaccharide than inducers of inflammation
Oskolkova OV, et al.
Journal of immunology (Baltimore, Md. : 1950), 185(12), 7706-7712 (2010)
Direct evidence that neural cell adhesion molecule (NCAM) polysialylation increases intermembrane repulsion and abrogates adhesion
Johnson CP, et al.
The Journal of Biological Chemistry, 280(1), 137-145 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service